Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561409_s1
          See other HTR1A GE Assays ›
          Gene Symbol:
          Htr1a
          Gene Name:
          5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled
          Gene Aliases:
          5HT1A, RAT5HT1A
          Chromosome Location:
          Chr.2: 36246628 - 36247896 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561409_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Htr1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012585.1 NP_036717.1 1 847 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Htr1a

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          cell proliferation
          regulation of serotonin secretion
          positive regulation of microtubule depolymerization
          exploration behavior
          regulation of dopamine metabolic process
          vasoconstriction
          serotonin metabolic process
          regulation of hormone secretion
          regulation of behavior
          regulation of feeding behavior
          NMDA glutamate receptor clustering
          G-protein alpha-subunit binding
          G-protein coupled serotonin receptor activity
          protein binding
          drug binding
          serotonin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052031 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052031 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052031 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812880 - -
          Affymetrix Rat Gene 1.1 ST Array 10812879 - -
          Rat Genome 230 2.0 Array 1369809_at - -
          Rat Genome 230 2.0 Array 1369809_PM_at - -
          Affymetrix HT Rat Focus Array 1369809_PM_at - -
          Affymetrix HT Rat Focus Array 1369809_at - -
          Affymetrix Rat Clariom S Assay PSR0200001843.rn.1 TC0200000318.rn.1 RTA TC
          Rat Expression Set 230 1369809_at - -
          Rat Expression Set 230 1369809_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812879 - -
          Rat Genome U34 Set J05276cds_at - -
          Rat Genome U34 Set J05276cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21370555 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372545 - -
          RatRef-12 Expression BeadChip ILMN_49989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline