Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561411_m1
          See other IAPP GE Assays ›
          Gene Symbol:
          Iapp
          Gene Name:
          islet amyloid polypeptide
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 176510203 - 176517903 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561411_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iapp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012586.2 NP_036718.1 1 - 2 83 71
          XM_006237614.2 XP_006237676.1 2 - 3 612 71
          GenBank mRNA J04544.1 - 2 - 3 170 71
          M25390.1 - 2 - 3 119 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002009.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Iapp

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          glucose metabolic process
          sensory perception of pain
          eating behavior
          negative regulation of cell differentiation
          negative regulation of bone resorption
          hormone activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125863 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003854 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003854 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003854 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6325 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859548 - -
          Affymetrix Rat Gene 1.0 ST Array 10859550 - -
          Affymetrix Rat Gene 1.0 ST Array 10859551 - -
          Affymetrix Rat Gene 1.0 ST Array 10859549 - -
          Affymetrix Rat Gene 1.1 ST Array 10859547 - -
          Rat Genome 230 2.0 Array 1387660_PM_at - -
          Rat Genome 230 2.0 Array 1387660_at - -
          Affymetrix HT Rat Focus Array 1387660_PM_at - -
          Affymetrix HT Rat Focus Array 1387660_at - -
          Affymetrix Rat Clariom S Assay PSR0400014482.rn.1 TC0400002009.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400014484.rn.1 TC0400002009.rn.1 RTA TC
          Rat Expression Set 230 1387660_at - -
          Rat Expression Set 230 1387660_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859547 - -
          Rat Genome U34 Set X52820mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20795859 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359967 - -
          RatRef-12 Expression BeadChip ILMN_61672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline