Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561502_m1
          See other ME1 GE Assays ›
          Gene Symbol:
          Me1
          Gene Name:
          malic enzyme 1, NADP(+)-dependent, cytosolic
          Gene Aliases:
          MOD1
          Chromosome Location:
          Chr.8: 94256830 - 94368834 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561502_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Me1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012600.2 NP_036732.2 9 - 10 1028 119
          XM_008766439.1 XP_008764661.1 9 - 10 1325 119
          GenBank mRNA CO560778.1 - 5 - 6 525 119
          FQ228526.1 - 9 - 10 1059 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003059.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Me1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          malate metabolic process
          response to hormone
          protein tetramerization
          oxidation-reduction process
          malic enzyme activity
          malate dehydrogenase (decarboxylating) (NAD+) activity
          malate dehydrogenase (decarboxylating) (NADP+) activity
          oxaloacetate decarboxylase activity
          manganese ion binding
          NADP binding
          NAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136153 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P792817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102933 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38939 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121519 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118486 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P792817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P792817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136153 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P792817 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P792817 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11481 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701024469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73526 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34853 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47406 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4069 - -
          Agilent Rat cDNA Microarray (G4105A) 701024469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919114 - -
          Affymetrix Rat Gene 1.0 ST Array 10919106 - -
          Affymetrix Rat Gene 1.0 ST Array 10919110 - -
          Affymetrix Rat Gene 1.0 ST Array 10919117 - -
          Affymetrix Rat Gene 1.0 ST Array 10919116 - -
          Affymetrix Rat Gene 1.0 ST Array 10919112 - -
          Affymetrix Rat Gene 1.0 ST Array 10919107 - -
          Affymetrix Rat Gene 1.0 ST Array 10919105 - -
          Affymetrix Rat Gene 1.0 ST Array 10919104 - -
          Affymetrix Rat Gene 1.0 ST Array 10919109 - -
          Affymetrix Rat Gene 1.0 ST Array 10919113 - -
          Affymetrix Rat Gene 1.0 ST Array 10919108 - -
          Affymetrix Rat Gene 1.0 ST Array 10919115 - -
          Affymetrix Rat Gene 1.0 ST Array 10919111 - -
          Affymetrix Rat Gene 1.1 ST Array 10919103 - -
          Rat Genome 230 2.0 Array 1370870_PM_at - -
          Rat Genome 230 2.0 Array 1370067_at - -
          Rat Genome 230 2.0 Array 1370067_PM_at - -
          Rat Genome 230 2.0 Array 1370870_at - -
          Affymetrix HT Rat Focus Array 1370870_at - -
          Affymetrix HT Rat Focus Array 1370067_at - -
          Affymetrix HT Rat Focus Array 1370067_PM_at - -
          Affymetrix HT Rat Focus Array 1370870_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800024550.rn.1 TC0800003059.rn.1 RTA TC
          Rat Expression Set 230 1370870_PM_at - -
          Rat Expression Set 230 1370870_at - -
          Rat Expression Set 230 1370067_at - -
          Rat Expression Set 230 1370067_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919103 - -
          Rat Genome U34 Set rc_AI171506_at - -
          Rat Genome U34 Set rc_AI171506_g_at - -
          Rat Genome U34 Set rc_AI008020_at - -
          Rat Genome U34 Set M26594_at - -
          Rat Genome U34 Set rc_AI171506_at - -
          Rat Genome U34 Set rc_AI171506_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21265914 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365946 - -
          RatRef-12 Expression BeadChip ILMN_53682 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline