Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561699_m1
          See other OPRD1 GE Assays ›
          Gene Symbol:
          Oprd1
          Gene Name:
          opioid receptor, delta 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 150288126 - 150323063 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561699_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oprd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012617.1 NP_036749.1 2 - 3 685 70
          GenBank mRNA D16348.1 - 2 - 3 685 70
          U00475.1 - 2 - 3 628 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003740.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oprd1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-activating dopamine receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          adult locomotory behavior
          negative regulation of gene expression
          negative regulation of protein oligomerization
          positive regulation of peptidyl-serine phosphorylation
          opioid receptor signaling pathway
          regulation of mitochondrial membrane potential
          regulation of calcium ion transport
          regulation of sensory perception of pain
          cellular response to growth factor stimulus
          cellular response to hypoxia
          cellular response to toxic substance
          opioid receptor activity
          enkephalin receptor activity
          neuropeptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P020053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145832 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020053 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P020053 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P020053 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P172990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880230 - -
          Affymetrix Rat Gene 1.0 ST Array 10880232 - -
          Affymetrix Rat Gene 1.0 ST Array 10880229 - -
          Affymetrix Rat Gene 1.0 ST Array 10880231 - -
          Affymetrix Rat Gene 1.1 ST Array 10880228 - -
          Rat Genome 230 2.0 Array 1368791_PM_at - -
          Rat Genome 230 2.0 Array 1368791_at - -
          Affymetrix HT Rat Focus Array 1368791_at - -
          Affymetrix HT Rat Focus Array 1368791_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500027899.rn.1 TC0500003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500027897.rn.1 TC0500003740.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500027898.rn.1 TC0500003740.rn.1 RTA TC
          Rat Expression Set 230 1368791_at - -
          Rat Expression Set 230 1368791_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880228 - -
          Rat Genome U34 Set U00475_at - -
          Rat Genome U34 Set U00475_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21771094 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361683 - -
          RatRef-12 Expression BeadChip ILMN_58002 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline