Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561900_m1
          See other SDC4 GE Assays ›
          Gene Symbol:
          Sdc4
          Gene Name:
          syndecan 4
          Gene Aliases:
          RATRYUDOCA, RYUDOCA
          Chromosome Location:
          Chr.3: 160872503 - 160891190 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012649.2 NP_036781.1 4 - 5 472 65
          GenBank mRNA M81786.1 - 4 - 5 472 65
          S61868.1 - 4 - 5 472 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004739.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdc4

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          cell adhesion
          signal transduction
          cell-cell signaling
          positive regulation of protein kinase activity
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          positive regulation of exosomal secretion
          positive regulation of extracellular exosome assembly
          fibronectin binding
          protein kinase C binding
          protein binding
          thrombospondin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P687186 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P090747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23181 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P687186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P687186 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P090747 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P090747 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P687186 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P687186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32384 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95958 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851604 - -
          Affymetrix Rat Gene 1.0 ST Array 10851605 - -
          Affymetrix Rat Gene 1.0 ST Array 10851600 - -
          Affymetrix Rat Gene 1.0 ST Array 10851606 - -
          Affymetrix Rat Gene 1.0 ST Array 10851601 - -
          Affymetrix Rat Gene 1.0 ST Array 10851603 - -
          Affymetrix Rat Gene 1.0 ST Array 10851602 - -
          Affymetrix Rat Gene 1.1 ST Array 10851599 - -
          Rat Genome 230 2.0 Array 1367721_PM_at - -
          Rat Genome 230 2.0 Array 1367721_at - -
          Affymetrix HT Rat Focus Array 1367721_at - -
          Affymetrix HT Rat Focus Array 1367721_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300036266.rn.1 TC0300004739.rn.1 RTA TC
          Rat Expression Set 230 1367721_at - -
          Rat Expression Set 230 1367721_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851599 - -
          Rat Genome U34 Set S61868_g_at - -
          Rat Genome U34 Set rc_AA800059_at - -
          Rat Genome U34 Set S61868_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22403998 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352387 - -
          RatRef-12 Expression BeadChip ILMN_60129 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline