Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561973_m1
          See other EEF1A2 GE Assays ›
          Gene Symbol:
          Eef1a2
          Gene Name:
          eukaryotic translation elongation factor 1 alpha 2
          Gene Aliases:
          Ps10, RATPS10, Stnl
          Chromosome Location:
          Chr.3: 176657104 - 176666282 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561973_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eef1a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012660.2 NP_036792.2 7 - 8 1592 82
          XM_006235735.2 XP_006235797.1 7 - 8 1410 82
          XM_008762467.1 XP_008760689.1 7 - 8 1364 82
          GenBank mRNA BC074016.1 - 7 - 8 1592 7105980 82
          M62751.1 - 7 - 8 1379 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005042.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eef1a2

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          response to inorganic substance
          positive regulation of apoptotic process
          positive regulation of lipid kinase activity
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31691 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099392 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099392 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099392 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1038178 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702028206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58870 - -
          Agilent Rat cDNA Microarray (G4105A) 702028206 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852465 - -
          Affymetrix Rat Gene 1.0 ST Array 10852467 - -
          Affymetrix Rat Gene 1.0 ST Array 10852471 - -
          Affymetrix Rat Gene 1.0 ST Array 10852470 - -
          Affymetrix Rat Gene 1.0 ST Array 10852473 - -
          Affymetrix Rat Gene 1.0 ST Array 10852468 - -
          Affymetrix Rat Gene 1.0 ST Array 10852472 - -
          Affymetrix Rat Gene 1.0 ST Array 10852469 - -
          Affymetrix Rat Gene 1.0 ST Array 10852466 - -
          Affymetrix Rat Gene 1.1 ST Array 10852464 - -
          Rat Genome 230 2.0 Array 1367799_at - -
          Rat Genome 230 2.0 Array 1395478_at - -
          Rat Genome 230 2.0 Array 1395478_PM_at - -
          Rat Genome 230 2.0 Array 1367799_PM_at - -
          Affymetrix HT Rat Focus Array 1367799_at - -
          Affymetrix HT Rat Focus Array 1395478_PM_at - -
          Affymetrix HT Rat Focus Array 1395478_at - -
          Affymetrix HT Rat Focus Array 1367799_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300038306.rn.1 TC0300005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038305.rn.1 TC0300005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038307.rn.1 TC0300005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038308.rn.1 TC0300005042.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038310.rn.1 TC0300005042.rn.1 RTA TC
          Rat Expression Set 230 1367799_at - -
          Rat Expression Set 230 1367799_PM_at - -
          Rat Expression Set 230 1395478_PM_at - -
          Rat Expression Set 230 1395478_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852464 - -
          Rat Genome U34 Set M62752_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21144815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367508 - -
          RatRef-12 Expression BeadChip ILMN_51222 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline