Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561981_m1
          See other STS GE Assays ›
          Gene Symbol:
          Sts
          Gene Name:
          steroid sulfatase (microsomal), isozyme S
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 45420469 - 45428748 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561981_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sts
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012661.1 NP_036793.1 9 - 10 1901 92
          XM_006256960.2 XP_006257022.1 9 - 10 2148 92
          XM_006256961.2 XP_006257023.1 9 - 10 2136 92
          XM_006256962.2 XP_006257024.1 9 - 10 1823 92
          GenBank mRNA BC085747.1 - 6 - 7 2577 92
          U37138.1 - 9 - 10 1901 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000430.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Androgen Panel Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sts

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          learning or memory
          metabolic process
          steroid metabolic process
          positive regulation of cell proliferation
          response to pH
          response to organic cyclic compound
          response to peptide hormone
          skin development
          response to estrogen
          steryl-sulfatase activity
          sulfuric ester hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P003589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118891 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003589 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P003589 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P003589 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P702550 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701486253 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702032882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98354 - -
          Agilent Rat cDNA Microarray (G4105A) 702032882 - -
          Agilent Rat cDNA Microarray (G4105A) 701486253 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933790 - -
          Affymetrix Rat Gene 1.0 ST Array 10933789 - -
          Affymetrix Rat Gene 1.0 ST Array 10933788 - -
          Affymetrix Rat Gene 1.0 ST Array 10933786 - -
          Affymetrix Rat Gene 1.0 ST Array 10933785 - -
          Affymetrix Rat Gene 1.0 ST Array 10933781 - -
          Affymetrix Rat Gene 1.0 ST Array 10933782 - -
          Affymetrix Rat Gene 1.0 ST Array 10933783 - -
          Affymetrix Rat Gene 1.0 ST Array 10933787 - -
          Affymetrix Rat Gene 1.0 ST Array 10933784 - -
          Affymetrix Rat Gene 1.1 ST Array 10933780 - -
          Rat Genome 230 2.0 Array 1368372_PM_at - -
          Rat Genome 230 2.0 Array 1368372_at - -
          Affymetrix HT Rat Focus Array 1368372_at - -
          Affymetrix HT Rat Focus Array 1368372_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002943.rn.1 TC0X00000430.rn.1 RTA TC
          Rat Expression Set 230 1368372_PM_at - -
          Rat Expression Set 230 1368372_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933780 - -
          Rat Genome U34 Set U37138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21376029 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368549 - -
          RatRef-12 Expression BeadChip ILMN_59743 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline