Search
Search
| Gene Symbol: Tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012668.2 | NP_036800.1 | 3 - 4 | 386 | 93 | |
| GenBank mRNA | BC089813.1 | - | 3 - 4 | 386 | 7372826 | 93 |
| M18340.1 | - | 3 - 4 | 439 | 93 | ||
| X02741.1 | - | 3 - 4 | 439 | 93 | ||
Set Membership: |
|