Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562048_m1
          See other THY1 GE Assays ›
          Gene Symbol:
          Thy1
          Gene Name:
          Thy-1 cell surface antigen
          Gene Aliases:
          CD7
          Chromosome Location:
          Chr.8: 48382121 - 48386935 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562048_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thy1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012673.2 NP_036805.1 2 - 3 130 74
          GenBank mRNA CB797376.1 - 2 - 3 130 74
          X03150.1 - 2 - 3 81 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000550.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thy1

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of protein kinase activity
          cytoskeleton organization
          single organismal cell-cell adhesion
          negative regulation of cell migration
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          mast cell activation
          retinal cone cell development
          focal adhesion assembly
          negative regulation of fibroblast proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of axonogenesis
          T cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of release of sequestered calcium ion into cytosol
          integrin binding
          protein binding
          enzyme binding
          protein kinase binding
          GPI anchor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P475159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475159 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P475159 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P475159 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10909409 - -
          Affymetrix Rat Gene 1.0 ST Array 10909410 - -
          Affymetrix Rat Gene 1.0 ST Array 10909408 - -
          Affymetrix Rat Gene 1.1 ST Array 10909407 - -
          Rat Genome 230 2.0 Array 1369652_PM_at - -
          Rat Genome 230 2.0 Array 1369652_at - -
          Rat Genome 230 2.0 Array 1369651_at - -
          Rat Genome 230 2.0 Array 1369651_PM_at - -
          Affymetrix HT Rat Focus Array 1369651_PM_at - -
          Affymetrix HT Rat Focus Array 1369652_at - -
          Affymetrix HT Rat Focus Array 1369651_at - -
          Affymetrix HT Rat Focus Array 1369652_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800003352.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003357.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003344.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003349.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003355.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003353.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003354.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003350.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003348.rn.1 TC0800000550.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800003345.rn.1 TC0800000550.rn.1 RTA TC
          Rat Expression Set 230 1369652_PM_at - -
          Rat Expression Set 230 1369652_at - -
          Rat Expression Set 230 1369651_PM_at - -
          Rat Expression Set 230 1369651_at - -
          Affymetrix Rat Gene 1.0 ST Array 10909407 - -
          Rat Genome U34 Set rc_AA874848_s_at - -
          Rat Genome U34 Set X02002_at - -
          Rat Genome U34 Set rc_AA851656_i_at - -
          Rat Genome U34 Set rc_AA851656_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22304317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367716 - -
          RatRef-12 Expression BeadChip ILMN_47878 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline