Search
Search
| Gene Symbol: Gcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012707.2 | NP_036839.2 | 2 - 3 | 154 | 65 | |
| GenBank mRNA | BP479145.1 | - | 2 - 3 | 154 | 65 | |
| BP480555.1 | - | 2 - 3 | 129 | 65 | ||
Set Membership: |
|