Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562332_m1
          See other SLC16A1 GE Assays ›
          Gene Symbol:
          Slc16a1
          Gene Name:
          solute carrier family 16 (monocarboxylate transporter), member 1
          Gene Aliases:
          MCT1, RATMCT1, RNMCT1
          Chromosome Location:
          Chr.2: 207108552 - 207128561 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562332_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc16a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012716.2 NP_036848.1 3 - 4 550 77
          GenBank mRNA BC078877.1 - 3 - 4 550 7122399 77
          D63834.1 - 564 77
          X86216.1 - 570 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001912.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc16a1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          response to food
          lactate transmembrane transport
          plasma membrane lactate transport
          glucose homeostasis
          regulation of insulin secretion
          centrosome organization
          behavioral response to nutrient
          cellular response to organic cyclic compound
          protein binding
          lactate transmembrane transporter activity
          symporter activity
          secondary active monocarboxylate transmembrane transporter activity
          protein homodimerization activity
          organic cyclic compound binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P086969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195693 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153354 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086969 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P086969 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P545426 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700253737 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52835 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76587 - -
          Agilent Rat cDNA Microarray (G4105A) 700253737 - -
          Agilent Rat cDNA Microarray (G4105A) 700462868 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818094 - -
          Affymetrix Rat Gene 1.0 ST Array 10818095 - -
          Affymetrix Rat Gene 1.0 ST Array 10818091 - -
          Affymetrix Rat Gene 1.0 ST Array 10818092 - -
          Affymetrix Rat Gene 1.0 ST Array 10818093 - -
          Affymetrix Rat Gene 1.0 ST Array 10818096 - -
          Affymetrix Rat Gene 1.1 ST Array 10818090 - -
          Rat Genome 230 2.0 Array 1380467_PM_at - -
          Rat Genome 230 2.0 Array 1397803_at - -
          Rat Genome 230 2.0 Array 1386981_at - -
          Rat Genome 230 2.0 Array 1380467_at - -
          Rat Genome 230 2.0 Array 1386981_PM_at - -
          Rat Genome 230 2.0 Array 1397803_PM_at - -
          Affymetrix HT Rat Focus Array 1380467_at - -
          Affymetrix HT Rat Focus Array 1386981_at - -
          Affymetrix HT Rat Focus Array 1386981_PM_at - -
          Affymetrix HT Rat Focus Array 1380467_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200013071.rn.1 TC0200001912.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200013074.rn.1 TC0200001912.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200013076.rn.1 TC0200001912.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200013077.rn.1 TC0200001912.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200013073.rn.1 TC0200001912.rn.1 RTA TC
          Rat Expression Set 230 1386981_at - -
          Rat Expression Set 230 1386981_PM_at - -
          Rat Expression Set 230 1380467_PM_at - -
          Rat Expression Set 230 1380467_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818090 - -
          Rat Genome U34 Set rc_AI145680_s_at - -
          Rat Genome U34 Set D63834_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21184793 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355191 - -
          RatRef-12 Expression BeadChip ILMN_55995 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline