Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562354_m1
          See other P2RX6 GE Assays ›
          Gene Symbol:
          P2rx6
          Gene Name:
          purinergic receptor P2X, ligand-gated ion channel, 6
          Gene Aliases:
          P2rxl1, P2x6, Rnap2x6
          Chromosome Location:
          Chr.11: 87434931 - 87445221 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562354_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P2rx6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012721.2 NP_036853.2 3 - 4 499 56
          XM_008768851.1 XP_008767073.1 3 - 4 689 56
          XM_008768852.1 XP_008767074.1 3 - 4 374 56
          GenBank mRNA X92070.1 - 3 - 4 380 56
          X97376.1 - 3 - 4 510 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001006.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P2rx6

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          ion transmembrane transport
          purinergic nucleotide receptor signaling pathway
          protein homooligomerization
          protein heterooligomerization
          cation transmembrane transport
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          ion channel activity
          protein binding
          ATP binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104998 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043259 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P367481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755701 - -
          Affymetrix Rat Gene 1.0 ST Array 10755696 - -
          Affymetrix Rat Gene 1.0 ST Array 10755702 - -
          Affymetrix Rat Gene 1.0 ST Array 10755704 - -
          Affymetrix Rat Gene 1.0 ST Array 10755697 - -
          Affymetrix Rat Gene 1.0 ST Array 10755695 - -
          Affymetrix Rat Gene 1.0 ST Array 10755703 - -
          Affymetrix Rat Gene 1.0 ST Array 10755694 - -
          Affymetrix Rat Gene 1.0 ST Array 10755699 - -
          Affymetrix Rat Gene 1.0 ST Array 10755698 - -
          Affymetrix Rat Gene 1.0 ST Array 10755700 - -
          Affymetrix Rat Gene 1.0 ST Array 10755693 - -
          Affymetrix Rat Gene 1.1 ST Array 10755692 - -
          Rat Genome 230 2.0 Array 1368740_PM_at - -
          Rat Genome 230 2.0 Array 1368740_at - -
          Affymetrix HT Rat Focus Array 1368740_at - -
          Affymetrix HT Rat Focus Array 1368740_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100005917.rn.1 TC1100001006.rn.1 RTA TC
          Rat Expression Set 230 1368740_PM_at - -
          Rat Expression Set 230 1368740_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755692 - -
          Rat Genome U34 Set X92070_at - -
          Rat Genome U34 Set X92070_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22148244 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369288 - -
          RatRef-12 Expression BeadChip ILMN_68585 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline