Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562362_m1
          See other PIGR GE Assays ›
          Gene Symbol:
          Pigr
          Gene Name:
          polymeric immunoglobulin receptor
          Gene Aliases:
          RNPIGR2, pIgA-R
          Chromosome Location:
          Chr.13: 47564176 - 47592093 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562362_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pigr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012723.4 NP_036855.3 568 99
          GenBank mRNA X15741.1 - 2 - 3 462 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000227.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pigr

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          retina homeostasis
          immunoglobulin transcytosis in epithelial cells mediated by polymeric immunoglobulin receptor
          epidermal growth factor receptor signaling pathway
          Fc receptor signaling pathway
          receptor clustering
          polymeric immunoglobulin receptor activity
          epidermal growth factor receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P510738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P510738 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P510738 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P510738 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P510738 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4977 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 285 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10763766 - -
          Affymetrix Rat Gene 1.0 ST Array 10763761 - -
          Affymetrix Rat Gene 1.0 ST Array 10763762 - -
          Affymetrix Rat Gene 1.0 ST Array 10763767 - -
          Affymetrix Rat Gene 1.0 ST Array 10763764 - -
          Affymetrix Rat Gene 1.0 ST Array 10763760 - -
          Affymetrix Rat Gene 1.0 ST Array 10763765 - -
          Affymetrix Rat Gene 1.0 ST Array 10763763 - -
          Affymetrix Rat Gene 1.0 ST Array 10763759 - -
          Affymetrix Rat Gene 1.1 ST Array 10763758 - -
          Rat Genome 230 2.0 Array 1371099_PM_at - -
          Rat Genome 230 2.0 Array 1371099_at - -
          Affymetrix Rat Clariom S Assay PSR1300001245.rn.1 TC1300000227.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10763758 - -
          Rat Genome U34 Set L13235UTR#1_f_at - -
          Rat Genome U34 Set U01145UTR#1_s_at - -
          Rat Genome U34 Set L14001UTR#1_s_at - -
          Rat Genome U34 Set L14003UTR#1_f_at - -
          Rat Genome U34 Set L13025UTR#1_f_at - -
          Rat Genome U34 Set L13237UTR#1_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21478589 - -
          ABI Rat Genome Survey Microarray 21761409 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367822 - -
          RatRef-12 Expression BeadChip ILMN_53019 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline