Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562369_m1
          See other FCER1A GE Assays ›
          Gene Symbol:
          Fcer1a
          Gene Name:
          Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
          Gene Aliases:
          Iger01, RATIGER01
          Chromosome Location:
          Chr.13: 91727253 - 91737106 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562369_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcer1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012724.4 NP_036856.1 2 - 3 379 64
          XM_008769780.1 XP_008768002.1 1 - 2 1084 64
          XM_008769781.1 XP_008768003.1 1 - 2 1084 64
          GenBank mRNA J03606.1 - 2 - 3 379 64
          M17153.1 - 2 - 3 383 64
          M21622.1 - 2 - 3 442 64
          M21623.1 - 2 - 3 409 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001967.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcer1a

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of type I hypersensitivity
          serotonin secretion
          immune response
          signal transduction
          cell surface receptor signaling pathway
          activation of JUN kinase activity
          leukotriene biosynthetic process
          Fc-epsilon receptor signaling pathway
          positive regulation of mast cell degranulation
          positive regulation of interleukin-3 biosynthetic process
          positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of calcium-mediated signaling
          IgE receptor activity
          high-affinity IgE receptor activity
          IgE binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P654682 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11261 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P654682 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052825 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P654682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P654682 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052825 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P461402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770061 - -
          Affymetrix Rat Gene 1.0 ST Array 10770060 - -
          Affymetrix Rat Gene 1.0 ST Array 10770058 - -
          Affymetrix Rat Gene 1.0 ST Array 10770059 - -
          Affymetrix Rat Gene 1.0 ST Array 10770064 - -
          Affymetrix Rat Gene 1.0 ST Array 10770062 - -
          Affymetrix Rat Gene 1.0 ST Array 10770063 - -
          Affymetrix Rat Gene 1.0 ST Array 10770065 - -
          Affymetrix Rat Gene 1.1 ST Array 10770057 - -
          Rat Genome 230 2.0 Array 1387713_PM_a_at - -
          Rat Genome 230 2.0 Array 1387713_a_at - -
          Affymetrix HT Rat Focus Array 1387713_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387713_a_at - -
          Affymetrix Rat Clariom S Assay PSR1300014137.rn.1 TC1300001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014142.rn.1 TC1300001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014136.rn.1 TC1300001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014138.rn.1 TC1300001967.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014143.rn.1 TC1300001967.rn.1 RTA TC
          Rat Expression Set 230 1387713_a_at - -
          Rat Expression Set 230 1387713_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770057 - -
          Rat Genome U34 Set M21622_at - -
          Rat Genome U34 Set M21622_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22121038 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364767 - -
          RatRef-12 Expression BeadChip ILMN_63949 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline