Search
Search
| Gene Symbol: Glp1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012728.1 | NP_036860.1 | 3 - 4 | 300 | 87 | |
| GenBank mRNA | M97797.1 | - | 3 - 4 | 300 | 87 | |
| S75952.1 | - | 3 - 4 | 294 | 87 | ||
Set Membership: |
|