Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562433_m1
          See other RBP1 GE Assays ›
          Gene Symbol:
          Rbp1
          Gene Name:
          retinol binding protein 1, cellular
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 106449321 - 106470842 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562433_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012733.4 NP_036865.1 2 - 3 348 77
          GenBank mRNA FQ209770.1 - 2 - 3 362 77
          FQ209927.1 - 2 - 3 332 77
          FQ210835.1 - 2 - 3 363 77
          FQ218090.1 - 2 - 3 362 77
          M16459.1 - 2 - 3 348 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbp1

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoic acid biosynthetic process
          transport
          regulation of granulocyte differentiation
          response to vitamin A
          retinol metabolic process
          retinoic acid metabolic process
          transporter activity
          retinal binding
          retinol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11537 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11537 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11537 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912443 - -
          Affymetrix Rat Gene 1.0 ST Array 10912441 - -
          Affymetrix Rat Gene 1.0 ST Array 10912442 - -
          Affymetrix Rat Gene 1.0 ST Array 10912440 - -
          Affymetrix Rat Gene 1.1 ST Array 10912439 - -
          Rat Genome 230 2.0 Array 1367939_PM_at - -
          Rat Genome 230 2.0 Array 1367939_at - -
          Affymetrix HT Rat Focus Array 1367939_at - -
          Affymetrix HT Rat Focus Array 1367939_PM_at - -
          Rat Expression Set 230 1367939_PM_at - -
          Rat Expression Set 230 1367939_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912439 - -
          Rat Genome U34 Set M19257_at - -
          Rat Genome U34 Set M19257_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21089198 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375320 - -
          RatRef-12 Expression BeadChip ILMN_70061 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline