Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562717_m1
          See other SP4 GE Assays ›
          Gene Symbol:
          Sp4
          Gene Name:
          Sp4 transcription factor
          Gene Aliases:
          HF-1b
          Chromosome Location:
          Chr.6: 146135877 - 146201344 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562717_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012761.1 NP_036893.1 4 - 5 2052 81
          GenBank mRNA U07610.1 - 4 - 5 2052 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003460.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sp4

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          regulation of heart contraction
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032479 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P147030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98840 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146170 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P147030 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P147030 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032479 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P147030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78491 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892787 - -
          Affymetrix Rat Gene 1.0 ST Array 10892791 - -
          Affymetrix Rat Gene 1.0 ST Array 10892788 - -
          Affymetrix Rat Gene 1.0 ST Array 10892790 - -
          Affymetrix Rat Gene 1.0 ST Array 10892792 - -
          Affymetrix Rat Gene 1.0 ST Array 10892789 - -
          Affymetrix Rat Gene 1.0 ST Array 10892793 - -
          Affymetrix Rat Gene 1.1 ST Array 10892786 - -
          Rat Genome 230 2.0 Array 1369363_PM_at - -
          Rat Genome 230 2.0 Array 1369363_at - -
          Affymetrix HT Rat Focus Array 1369363_PM_at - -
          Affymetrix HT Rat Focus Array 1369363_at - -
          Affymetrix Rat Clariom S Assay PSR0600023810.rn.1 TC0600003460.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023809.rn.1 TC0600003460.rn.1 RTA TC
          Rat Expression Set 230 1369363_PM_at - -
          Rat Expression Set 230 1369363_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892786 - -
          Rat Genome U34 Set U07610_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21688749 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376532 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline