Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562775_m1
          See other GUCY1B3 GE Assays ›
          Gene Symbol:
          Gucy1b3
          Gene Name:
          guanylate cyclase 1, soluble, beta 3
          Gene Aliases:
          Gucy1b1, SGC
          Chromosome Location:
          Chr.2: 180976939 - 181026001 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562775_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gucy1b3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012769.2 NP_036901.2 3 - 4 269 135
          GenBank mRNA AB098025.1 - 2 - 3 181 135
          AB098707.1 - 2 - 3 181 135
          AB099521.2 - 2 - 3 181 135
          BC081840.1 - 3 - 4 269 7104600 135
          M22562.1 - 3 - 4 222 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003929.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gucy1b3

          Panther Classification:

          Molecular Function -

          guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          nitric oxide mediated signal transduction
          guanylate cyclase activity
          protein binding
          GTP binding
          heme binding
          ion binding
          metal ion binding
          protein heterodimerization activity
          Hsp90 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P535268 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P126685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77542 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159244 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25575 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535268 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126685 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535268 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P535268 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P126685 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P535268 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700521408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18470 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99874 - -
          Agilent Rat cDNA Microarray (G4105A) 700521408 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823906 - -
          Affymetrix Rat Gene 1.0 ST Array 10823910 - -
          Affymetrix Rat Gene 1.0 ST Array 10823916 - -
          Affymetrix Rat Gene 1.0 ST Array 10823917 - -
          Affymetrix Rat Gene 1.0 ST Array 10823907 - -
          Affymetrix Rat Gene 1.0 ST Array 10823919 - -
          Affymetrix Rat Gene 1.0 ST Array 10823913 - -
          Affymetrix Rat Gene 1.0 ST Array 10823918 - -
          Affymetrix Rat Gene 1.0 ST Array 10823911 - -
          Affymetrix Rat Gene 1.0 ST Array 10823908 - -
          Affymetrix Rat Gene 1.0 ST Array 10823912 - -
          Affymetrix Rat Gene 1.0 ST Array 10823914 - -
          Affymetrix Rat Gene 1.0 ST Array 10823905 - -
          Affymetrix Rat Gene 1.0 ST Array 10823904 - -
          Affymetrix Rat Gene 1.0 ST Array 10823909 - -
          Affymetrix Rat Gene 1.0 ST Array 10823915 - -
          Affymetrix Rat Gene 1.1 ST Array 10823903 - -
          Rat Genome 230 2.0 Array 1369097_PM_s_at - -
          Rat Genome 230 2.0 Array 1374389_at - -
          Rat Genome 230 2.0 Array 1374389_PM_at - -
          Rat Genome 230 2.0 Array 1369097_s_at - -
          Affymetrix HT Rat Focus Array 1374389_PM_at - -
          Affymetrix HT Rat Focus Array 1369097_s_at - -
          Affymetrix HT Rat Focus Array 1374389_at - -
          Affymetrix HT Rat Focus Array 1369097_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0200026375.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026379.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026383.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026378.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026370.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026371.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026380.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026372.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026374.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026376.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026373.rn.1 TC0200003929.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200026381.rn.1 TC0200003929.rn.1 RTA TC
          Rat Expression Set 230 1369097_PM_s_at - -
          Rat Expression Set 230 1369097_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823903 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21553475 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371639 - -
          RatRef-12 Expression BeadChip ILMN_57719 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline