Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562822_m1
          See other ADRBK1 GE Assays ›
          Gene Symbol:
          Adrbk1
          Gene Name:
          adrenergic, beta, receptor kinase 1
          Gene Aliases:
          BARK1, GRK-2, Grk2
          Chromosome Location:
          Chr.1: 219536220 - 219544329 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562822_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adrbk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012776.1 NP_036908.1 261 82
          GenBank mRNA M87854.1 - 261 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007069.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adrbk1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          desensitization of G-protein coupled receptor protein signaling pathway
          negative regulation of the force of heart contraction by chemical signal
          protein phosphorylation
          response to oxidative stress
          G-protein coupled receptor signaling pathway
          tachykinin receptor signaling pathway
          heart development
          aging
          response to organic cyclic compound
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          receptor internalization
          positive regulation of catecholamine secretion
          termination of G-protein coupled receptor signaling pathway
          response to hydrogen peroxide
          follicle-stimulating hormone signaling pathway
          negative regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of striated muscle contraction
          cardiac muscle contraction
          protein kinase activity
          protein serine/threonine kinase activity
          G-protein coupled receptor kinase activity
          protein binding
          ATP binding
          Edg-2 lysophosphatidic acid receptor binding
          beta-adrenergic receptor kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P491621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163589 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P491621 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P491621 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491621 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700292421 - -
          Agilent Rat cDNA Microarray (G4105A) 700292421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727549 - -
          Affymetrix Rat Gene 1.0 ST Array 10727554 - -
          Affymetrix Rat Gene 1.0 ST Array 10727555 - -
          Affymetrix Rat Gene 1.0 ST Array 10727551 - -
          Affymetrix Rat Gene 1.0 ST Array 10727540 - -
          Affymetrix Rat Gene 1.0 ST Array 10727547 - -
          Affymetrix Rat Gene 1.0 ST Array 10727539 - -
          Affymetrix Rat Gene 1.0 ST Array 10727541 - -
          Affymetrix Rat Gene 1.0 ST Array 10727546 - -
          Affymetrix Rat Gene 1.0 ST Array 10727557 - -
          Affymetrix Rat Gene 1.0 ST Array 10727552 - -
          Affymetrix Rat Gene 1.0 ST Array 10727538 - -
          Affymetrix Rat Gene 1.0 ST Array 10727553 - -
          Affymetrix Rat Gene 1.0 ST Array 10727556 - -
          Affymetrix Rat Gene 1.0 ST Array 10727550 - -
          Affymetrix Rat Gene 1.0 ST Array 10727544 - -
          Affymetrix Rat Gene 1.0 ST Array 10727543 - -
          Affymetrix Rat Gene 1.0 ST Array 10727542 - -
          Affymetrix Rat Gene 1.0 ST Array 10727545 - -
          Affymetrix Rat Gene 1.0 ST Array 10727548 - -
          Affymetrix Rat Gene 1.1 ST Array 10727537 - -
          Rat Genome 230 2.0 Array 1388757_PM_at - -
          Rat Genome 230 2.0 Array 1387429_at - -
          Rat Genome 230 2.0 Array 1388757_at - -
          Rat Genome 230 2.0 Array 1387429_PM_at - -
          Affymetrix HT Rat Focus Array 1388757_PM_at - -
          Affymetrix HT Rat Focus Array 1387429_PM_at - -
          Affymetrix HT Rat Focus Array 1388757_at - -
          Affymetrix HT Rat Focus Array 1387429_at - -
          Affymetrix Rat Clariom S Assay PSR0100056275.rn.1 TC0100007069.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100056273.rn.1 TC0100007069.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100056272.rn.1 TC0100007069.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100056274.rn.1 TC0100007069.rn.1 RTA TC
          Rat Expression Set 230 1387429_at - -
          Rat Expression Set 230 1387429_PM_at - -
          Rat Expression Set 230 1388757_at - -
          Rat Expression Set 230 1388757_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727537 - -
          Rat Genome U34 Set S48813_s_at - -
          Rat Genome U34 Set M87854_at - -
          Rat Genome U34 Set rc_AA998430_at - -
          Rat Genome U34 Set S48813_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21895880 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376677 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline