Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562940_m1
          See other DYRK1A GE Assays ›
          Gene Symbol:
          Dyrk1a
          Gene Name:
          dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A
          Gene Aliases:
          Dyrk, PSK47
          Chromosome Location:
          Chr.11: 34858340 - 34958733 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562940_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dyrk1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012791.2 NP_036923.1 5 - 6 637 86
          XM_006248031.2 XP_006248093.1 5 - 6 3582 86
          XM_008768562.1 XP_008766784.1 4 - 5 2845 86
          XM_008768563.1 XP_008766785.1 6 - 7 3718 86
          XM_008768564.1 XP_008766786.1 5 - 6 684 86
          XM_008768565.1 XP_008766787.1 5 - 6 759 86
          GenBank mRNA X79769.1 - 5 - 6 639 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000329.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dyrk1a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          protein phosphorylation
          nervous system development
          circadian rhythm
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          peptidyl-tyrosine phosphorylation
          regulation of endocytosis
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          protein autophosphorylation
          negative regulation of mRNA splicing, via spliceosome
          positive regulation of protein deacetylation
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          ATP binding
          identical protein binding
          protein self-association
          tau protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P514244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149409 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2811 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150482 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P514244 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P514244 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P514244 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P514244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92877 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14138 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45030 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750448 - -
          Affymetrix Rat Gene 1.0 ST Array 10750447 - -
          Affymetrix Rat Gene 1.0 ST Array 10750439 - -
          Affymetrix Rat Gene 1.0 ST Array 10750443 - -
          Affymetrix Rat Gene 1.0 ST Array 10750451 - -
          Affymetrix Rat Gene 1.0 ST Array 10750445 - -
          Affymetrix Rat Gene 1.0 ST Array 10750442 - -
          Affymetrix Rat Gene 1.0 ST Array 10750444 - -
          Affymetrix Rat Gene 1.0 ST Array 10750450 - -
          Affymetrix Rat Gene 1.0 ST Array 10750440 - -
          Affymetrix Rat Gene 1.0 ST Array 10750446 - -
          Affymetrix Rat Gene 1.0 ST Array 10750441 - -
          Affymetrix Rat Gene 1.0 ST Array 10750438 - -
          Affymetrix Rat Gene 1.1 ST Array 10750437 - -
          Rat Genome 230 2.0 Array 1370183_at - -
          Rat Genome 230 2.0 Array 1370183_PM_at - -
          Affymetrix HT Rat Focus Array 1370183_at - -
          Affymetrix HT Rat Focus Array 1370183_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100001586.rn.1 TC1100000329.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10750437 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21922689 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372137 - -
          RatRef-12 Expression BeadChip ILMN_48344 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline