Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562985_s1
          See other ID1 GE Assays ›
          Gene Symbol:
          Id1
          Gene Name:
          inhibitor of DNA binding 1
          Gene Aliases:
          ID125A, Idb1
          Chromosome Location:
          Chr.3: 148214622 - 148216715 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562985_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Id1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012797.2 NP_036929.2 1 351 75
          XM_006235267.2 XP_006235329.1 1 1317 75
          GenBank mRNA BC127448.1 - 1 363 8372348 75
          D10862.1 - 1 332 75
          L23148.1 - 1 352 75
          M86708.1 - 1 358 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Id1

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          endothelial cell morphogenesis
          transcription, DNA-templated
          brain development
          heart development
          circadian rhythm
          positive regulation of gene expression
          response to organic cyclic compound
          neuron differentiation
          BMP signaling pathway
          protein destabilization
          negative regulation of protein binding
          positive regulation of actin filament bundle assembly
          collagen metabolic process
          cell-abiotic substrate adhesion
          negative regulation of apoptotic process
          negative regulation of DNA binding
          regulation of MAPK cascade
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of endothelial cell differentiation
          negative regulation of osteoblast differentiation
          regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          response to antibiotic
          positive regulation of epithelial cell proliferation
          negative regulation of dendrite morphogenesis
          lung morphogenesis
          lung vasculature development
          cellular response to epidermal growth factor stimulus
          cellular response to transforming growth factor beta stimulus
          regulation of vasculature development
          cellular response to peptide
          negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          cellular response to dopamine
          cellular response to nerve growth factor stimulus
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein C-terminus binding
          transcription factor binding
          protein homodimerization activity
          protein N-terminus binding
          proteasome binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P723173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634395 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P723173 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P723173 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P723173 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P723173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25581 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840894 - -
          Affymetrix Rat Gene 1.0 ST Array 10840892 - -
          Affymetrix Rat Gene 1.0 ST Array 10840891 - -
          Affymetrix Rat Gene 1.0 ST Array 10840893 - -
          Affymetrix Rat Gene 1.1 ST Array 10840890 - -
          Rat Genome 230 2.0 Array 1387028_a_at - -
          Rat Genome 230 2.0 Array 1387028_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387028_a_at - -
          Affymetrix HT Rat Focus Array 1387028_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300014623.rn.1 TC0300002046.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014622.rn.1 TC0300002046.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014619.rn.1 TC0300002046.rn.1 RTA TC
          Rat Expression Set 230 1387028_a_at - -
          Rat Expression Set 230 1387028_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840890 - -
          Rat Genome U34 Set L23148_g_at - -
          Rat Genome U34 Set L23148_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21389601 - -
          ABI Rat Genome Survey Microarray 22259866 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365871 - -
          RatRef-12 Expression BeadChip ILMN_56825 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline