Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563013_m1
          See other ABCD3 GE Assays ›
          Gene Symbol:
          Abcd3
          Gene Name:
          ATP-binding cassette, subfamily D (ALD), member 3
          Gene Aliases:
          PMP70, Pxmp1
          Chromosome Location:
          Chr.2: 225335708 - 225389120 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563013_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012804.1 NP_036936.1 20 - 21 1772 87
          GenBank mRNA D90038.1 - 20 - 21 1772 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcd3

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          peroxisome organization
          metabolic process
          response to organic cyclic compound
          peroxisomal long-chain fatty acid import
          response to drug
          very long-chain fatty acid catabolic process
          transmembrane transport
          protein binding
          ATP binding
          ATPase activity
          ATPase activity, coupled to transmembrane movement of substances
          protein homodimerization activity
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11570 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11570 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11570 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11570 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701427356 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700288719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101859 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54793 - -
          Agilent Rat cDNA Microarray (G4105A) 701427356 - -
          Agilent Rat cDNA Microarray (G4105A) 700288719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10826495 - -
          Affymetrix Rat Gene 1.0 ST Array 10826482 - -
          Affymetrix Rat Gene 1.0 ST Array 10826488 - -
          Affymetrix Rat Gene 1.0 ST Array 10826485 - -
          Affymetrix Rat Gene 1.0 ST Array 10826480 - -
          Affymetrix Rat Gene 1.0 ST Array 10826498 - -
          Affymetrix Rat Gene 1.0 ST Array 10826484 - -
          Affymetrix Rat Gene 1.0 ST Array 10826493 - -
          Affymetrix Rat Gene 1.0 ST Array 10826476 - -
          Affymetrix Rat Gene 1.0 ST Array 10826483 - -
          Affymetrix Rat Gene 1.0 ST Array 10826490 - -
          Affymetrix Rat Gene 1.0 ST Array 10826489 - -
          Affymetrix Rat Gene 1.0 ST Array 10826487 - -
          Affymetrix Rat Gene 1.0 ST Array 10826494 - -
          Affymetrix Rat Gene 1.0 ST Array 10826492 - -
          Affymetrix Rat Gene 1.0 ST Array 10826478 - -
          Affymetrix Rat Gene 1.0 ST Array 10826477 - -
          Affymetrix Rat Gene 1.0 ST Array 10826499 - -
          Affymetrix Rat Gene 1.0 ST Array 10826486 - -
          Affymetrix Rat Gene 1.0 ST Array 10826496 - -
          Affymetrix Rat Gene 1.0 ST Array 10826481 - -
          Affymetrix Rat Gene 1.0 ST Array 10826497 - -
          Affymetrix Rat Gene 1.0 ST Array 10826479 - -
          Affymetrix Rat Gene 1.1 ST Array 10826474 - -
          Rat Genome 230 2.0 Array 1368057_PM_at - -
          Rat Genome 230 2.0 Array 1368057_at - -
          Affymetrix HT Rat Focus Array 1368057_PM_at - -
          Affymetrix HT Rat Focus Array 1368057_at - -
          Affymetrix Rat Clariom S Assay PSR0200031934.rn.1 TC0200004587.rn.1 RTA TC
          Rat Expression Set 230 1368057_at - -
          Rat Expression Set 230 1368057_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10826474 - -
          Rat Genome U34 Set D90038_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20914137 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357729 - -
          RatRef-12 Expression BeadChip ILMN_51315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0