Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563116_m1
          See other IGFBP5 GE Assays ›
          Gene Symbol:
          Igfbp5
          Gene Name:
          insulin-like growth factor binding protein 5
          Gene Aliases:
          IGF-BP5
          Chromosome Location:
          Chr.9: 80154230 - 80166813 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563116_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igfbp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012817.1 NP_036949.1 3 - 4 1233 58
          GenBank mRNA BC087030.1 - 1435 58
          M62781.1 - 3 - 4 1233 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002215.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igfbp5

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          osteoblast differentiation
          glucose metabolic process
          regulation of glucose metabolic process
          negative regulation of smooth muscle cell migration
          negative regulation of translation
          negative regulation of cell migration
          hair follicle morphogenesis
          intracellular signal transduction
          regulation of growth
          glucose homeostasis
          regulation of insulin-like growth factor receptor signaling pathway
          positive regulation of insulin-like growth factor receptor signaling pathway
          negative regulation of insulin-like growth factor receptor signaling pathway
          type B pancreatic cell proliferation
          negative regulation of osteoblast differentiation
          skeletal muscle tissue growth
          negative regulation of smooth muscle cell proliferation
          striated muscle cell differentiation
          positive regulation of protein kinase B signaling
          mammary gland involution
          response to growth hormone
          cellular response to cAMP
          cellular response to organic cyclic compound
          fibronectin binding
          insulin-like growth factor binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047478 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P264240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047478 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P264240 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047478 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P285534 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264240 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700145350 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102108 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59089 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92619 - -
          Agilent Rat cDNA Microarray (G4105A) 700145350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928843 - -
          Affymetrix Rat Gene 1.0 ST Array 10928840 - -
          Affymetrix Rat Gene 1.0 ST Array 10928839 - -
          Affymetrix Rat Gene 1.0 ST Array 10928841 - -
          Affymetrix Rat Gene 1.0 ST Array 10928842 - -
          Affymetrix Rat Gene 1.0 ST Array 10928838 - -
          Affymetrix Rat Gene 1.1 ST Array 10928837 - -
          Rat Genome 230 2.0 Array 1387348_PM_at - -
          Rat Genome 230 2.0 Array 1397830_at - -
          Rat Genome 230 2.0 Array 1396152_s_at - -
          Rat Genome 230 2.0 Array 1387348_at - -
          Rat Genome 230 2.0 Array 1387347_PM_at - -
          Rat Genome 230 2.0 Array 1370960_PM_at - -
          Rat Genome 230 2.0 Array 1387347_at - -
          Rat Genome 230 2.0 Array 1397830_PM_at - -
          Rat Genome 230 2.0 Array 1370960_at - -
          Rat Genome 230 2.0 Array 1396152_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387348_at - -
          Affymetrix HT Rat Focus Array 1387347_PM_at - -
          Affymetrix HT Rat Focus Array 1387347_at - -
          Affymetrix HT Rat Focus Array 1370960_at - -
          Affymetrix HT Rat Focus Array 1387348_PM_at - -
          Affymetrix HT Rat Focus Array 1396152_PM_s_at - -
          Affymetrix HT Rat Focus Array 1370960_PM_at - -
          Affymetrix HT Rat Focus Array 1396152_s_at - -
          Affymetrix Rat Clariom S Assay PSR0900018019.rn.1 TC0900002215.rn.1 RTA TC
          Rat Expression Set 230 1370960_PM_at - -
          Rat Expression Set 230 1387348_at - -
          Rat Expression Set 230 1387347_at - -
          Rat Expression Set 230 1370960_at - -
          Rat Expression Set 230 1387347_PM_at - -
          Rat Expression Set 230 1387348_PM_at - -
          Rat Expression Set 230 1396152_s_at - -
          Rat Expression Set 230 1397830_PM_at - -
          Rat Expression Set 230 1396152_PM_s_at - -
          Rat Expression Set 230 1397830_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928837 - -
          Rat Genome U34 Set rc_AI029920_s_at - -
          Rat Genome U34 Set M62781_at - -
          Rat Genome U34 Set rc_AI070577_at - -
          Rat Genome U34 Set rc_AI176584_at - -
          Rat Genome U34 Set rc_AI029920_s_at - -
          Rat Genome U34 Set M62781_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20755729 - -
          ABI Rat Genome Survey Microarray 22374512 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369802 - -
          RatRef-12 Expression BeadChip ILMN_67540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline