Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563121_m1
          See other ACADL GE Assays ›
          Gene Symbol:
          Acadl
          Gene Name:
          acyl-CoA dehydrogenase, long chain
          Gene Aliases:
          ACOADA, LCAD
          Chromosome Location:
          Chr.9: 73833368 - 73871857 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563121_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acadl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012819.1 NP_036951.1 1 - 2 82 74
          GenBank mRNA BC062006.1 - 1 - 2 80 5599828 74
          FQ215575.1 - 1 - 2 133 74
          J05029.1 - 1 - 2 93 74
          L11276.1 - 1 - 2 82 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002139.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acadl

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          carnitine metabolic process, CoA-linked
          fatty acid beta-oxidation using acyl-CoA dehydrogenase
          carnitine catabolic process
          long-chain fatty acid catabolic process
          cellular lipid catabolic process
          negative regulation of fatty acid biosynthetic process
          negative regulation of fatty acid oxidation
          protein homotetramerization
          oxidation-reduction process
          regulation of cholesterol metabolic process
          fatty-acyl-CoA binding
          long-chain-acyl-CoA dehydrogenase activity
          palmitoyl-CoA oxidase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93855 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14184 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14184 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14184 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700182615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23707 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66870 - -
          Agilent Rat cDNA Microarray (G4105A) 700182615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10928603 - -
          Affymetrix Rat Gene 1.0 ST Array 10928613 - -
          Affymetrix Rat Gene 1.0 ST Array 10928612 - -
          Affymetrix Rat Gene 1.0 ST Array 10928606 - -
          Affymetrix Rat Gene 1.0 ST Array 10928608 - -
          Affymetrix Rat Gene 1.0 ST Array 10928604 - -
          Affymetrix Rat Gene 1.0 ST Array 10928609 - -
          Affymetrix Rat Gene 1.0 ST Array 10928605 - -
          Affymetrix Rat Gene 1.0 ST Array 10928607 - -
          Affymetrix Rat Gene 1.0 ST Array 10928611 - -
          Affymetrix Rat Gene 1.0 ST Array 10928610 - -
          Affymetrix Rat Gene 1.1 ST Array 10928602 - -
          Rat Genome 230 2.0 Array 1367735_at - -
          Rat Genome 230 2.0 Array 1367735_PM_at - -
          Affymetrix HT Rat Focus Array 1367735_PM_at - -
          Affymetrix HT Rat Focus Array 1367735_at - -
          Affymetrix Rat Clariom S Assay PSR0900017586.rn.1 TC0900002139.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900017585.rn.1 TC0900002139.rn.1 RTA TC
          Rat Expression Set 230 1367735_at - -
          Rat Expression Set 230 1367735_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10928602 - -
          Rat Genome U34 Set J05029_s_at - -
          Rat Genome U34 Set rc_AA848753_at - -
          Rat Genome U34 Set J05029_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21359296 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355733 - -
          RatRef-12 Expression BeadChip ILMN_69215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline