Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563162_m1
          See other ADCY6 GE Assays ›
          Gene Symbol:
          Adcy6
          Gene Name:
          adenylate cyclase 6
          Gene Aliases:
          ACVI, ADCYB
          Chromosome Location:
          Chr.7: 140270692 - 140291620 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563162_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adcy6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270785.1 NP_001257714.1 2 - 3 914 58
          GenBank mRNA L01115.1 - 2 - 3 1057 58
          M96160.1 - 2 - 3 914 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003853.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP biosynthetic process
          dopamine receptor signaling pathway
          negative regulation of neuron projection development
          intracellular signal transduction
          cellular response to prostaglandin E stimulus
          cellular response to catecholamine stimulus
          maintenance of protein location in plasma membrane
          SNARE binding
          nucleotide binding
          adenylate cyclase activity
          receptor binding
          ATP binding
          calcium- and calmodulin-responsive adenylate cyclase activity
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139493 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173004 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11576 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P107300 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700367185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47234 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32728 - -
          Agilent Rat cDNA Microarray (G4105A) 700367185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906892 - -
          Affymetrix Rat Gene 1.0 ST Array 10906905 - -
          Affymetrix Rat Gene 1.0 ST Array 10906891 - -
          Affymetrix Rat Gene 1.0 ST Array 10906887 - -
          Affymetrix Rat Gene 1.0 ST Array 10906893 - -
          Affymetrix Rat Gene 1.0 ST Array 10906903 - -
          Affymetrix Rat Gene 1.0 ST Array 10906896 - -
          Affymetrix Rat Gene 1.0 ST Array 10906895 - -
          Affymetrix Rat Gene 1.0 ST Array 10906902 - -
          Affymetrix Rat Gene 1.0 ST Array 10906885 - -
          Affymetrix Rat Gene 1.0 ST Array 10906890 - -
          Affymetrix Rat Gene 1.0 ST Array 10906884 - -
          Affymetrix Rat Gene 1.0 ST Array 10906889 - -
          Affymetrix Rat Gene 1.0 ST Array 10906894 - -
          Affymetrix Rat Gene 1.0 ST Array 10906883 - -
          Affymetrix Rat Gene 1.0 ST Array 10906904 - -
          Affymetrix Rat Gene 1.0 ST Array 10906900 - -
          Affymetrix Rat Gene 1.0 ST Array 10906897 - -
          Affymetrix Rat Gene 1.0 ST Array 10906886 - -
          Affymetrix Rat Gene 1.0 ST Array 10906901 - -
          Affymetrix Rat Gene 1.0 ST Array 10906888 - -
          Affymetrix Rat Gene 1.0 ST Array 10906899 - -
          Affymetrix Rat Gene 1.0 ST Array 10906898 - -
          Affymetrix Rat Gene 1.1 ST Array 10906882 - -
          Rat Genome 230 2.0 Array 1367644_at - -
          Rat Genome 230 2.0 Array 1367644_PM_at - -
          Rat Genome 230 2.0 Array 1386876_PM_at - -
          Rat Genome 230 2.0 Array 1386876_at - -
          Affymetrix HT Rat Focus Array 1386876_PM_at - -
          Affymetrix HT Rat Focus Array 1367644_PM_at - -
          Affymetrix HT Rat Focus Array 1367644_at - -
          Affymetrix HT Rat Focus Array 1386876_at - -
          Affymetrix Rat Clariom S Assay PSR0700030878.rn.1 TC0700003853.rn.1 RTA TC
          Rat Expression Set 230 1386876_PM_at - -
          Rat Expression Set 230 1367644_PM_at - -
          Rat Expression Set 230 1367644_at - -
          Rat Expression Set 230 1386876_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906882 - -
          Rat Genome U34 Set L01115_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21789873 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362372 - -
          RatRef-12 Expression BeadChip ILMN_58261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline