Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563345_m1
          See other EMP1 GE Assays ›
          Gene Symbol:
          Emp1
          Gene Name:
          epithelial membrane protein 1
          Gene Aliases:
          CL-20, EMP-1, ENP1MR, TMP
          Chromosome Location:
          Chr.4: 169161072 - 169181967 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563345_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012843.2 NP_036975.1 222 63
          XM_006237515.2 XP_006237577.1 331 63
          XM_008763369.1 XP_008761591.1 331 63
          GenBank mRNA BC087031.1 - 222 7192247 63
          Z54212.1 - 191 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001941.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emp1

          Panther Classification:

          Molecular Function -

          cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          cell death
          cell growth
          bleb assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P807058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P807058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P807058 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P807058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P807058 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17255 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859298 - -
          Affymetrix Rat Gene 1.0 ST Array 10859301 - -
          Affymetrix Rat Gene 1.0 ST Array 10859299 - -
          Affymetrix Rat Gene 1.0 ST Array 10859300 - -
          Affymetrix Rat Gene 1.0 ST Array 10859297 - -
          Affymetrix Rat Gene 1.1 ST Array 10859296 - -
          Rat Genome 230 2.0 Array 1371527_at - -
          Rat Genome 230 2.0 Array 1369736_at - -
          Rat Genome 230 2.0 Array 1371527_PM_at - -
          Rat Genome 230 2.0 Array 1369736_PM_at - -
          Affymetrix HT Rat Focus Array 1371527_PM_at - -
          Affymetrix HT Rat Focus Array 1369736_at - -
          Affymetrix HT Rat Focus Array 1371527_at - -
          Affymetrix HT Rat Focus Array 1369736_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400014003.rn.1 TC0400001941.rn.1 RTA TC
          Rat Expression Set 230 1371527_PM_at - -
          Rat Expression Set 230 1369736_PM_at - -
          Rat Expression Set 230 1371527_at - -
          Rat Expression Set 230 1369736_at - -
          Affymetrix Rat Gene 1.0 ST Array 10859296 - -
          Rat Genome U34 Set Z54212_at - -
          Rat Genome U34 Set rc_AI070336_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21226588 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350180 - -
          RatRef-12 Expression BeadChip ILMN_48170 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline