Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563377_m1
          See other GHRHR GE Assays ›
          Gene Symbol:
          Ghrhr
          Gene Name:
          growth hormone releasing hormone receptor
          Gene Aliases:
          GHRHREC
          Chromosome Location:
          Chr.4: 85587321 - 85601666 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563377_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ghrhr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012850.1 NP_036982.1 1 - 2 84 95
          XM_008762872.1 XP_008761094.1 1 - 2 1447 95
          GenBank mRNA AF122055.1 - 1 - 2 57 95
          L01407.1 - 1 - 2 84 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000975.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ghrhr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          lactation
          positive regulation of cell proliferation
          determination of adult lifespan
          cAMP-mediated signaling
          adenohypophysis development
          water homeostasis
          growth hormone secretion
          positive regulation of cAMP metabolic process
          positive regulation of cAMP biosynthetic process
          mammary gland development
          response to insulin
          cellular response to insulin stimulus
          regulation of intracellular steroid hormone receptor signaling pathway
          regulation of growth
          positive regulation of multicellular organism growth
          hormone metabolic process
          regulation of insulin-like growth factor receptor signaling pathway
          response to estrogen
          positive regulation of hormone secretion
          cell maturation
          regulation of protein metabolic process
          response to glucocorticoid
          positive regulation of growth hormone secretion
          somatotropin secreting cell development
          growth hormone-releasing hormone receptor activity
          peptide hormone binding
          growth factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11586 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P145142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20448 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1560 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145142 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P145142 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P145142 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11586 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292462 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702075474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57747 - -
          Agilent Rat cDNA Microarray (G4105A) 702075474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855717 - -
          Affymetrix Rat Gene 1.0 ST Array 10855712 - -
          Affymetrix Rat Gene 1.0 ST Array 10855711 - -
          Affymetrix Rat Gene 1.0 ST Array 10855713 - -
          Affymetrix Rat Gene 1.0 ST Array 10855721 - -
          Affymetrix Rat Gene 1.0 ST Array 10855710 - -
          Affymetrix Rat Gene 1.0 ST Array 10855709 - -
          Affymetrix Rat Gene 1.0 ST Array 10855714 - -
          Affymetrix Rat Gene 1.0 ST Array 10855723 - -
          Affymetrix Rat Gene 1.0 ST Array 10855722 - -
          Affymetrix Rat Gene 1.0 ST Array 10855724 - -
          Affymetrix Rat Gene 1.0 ST Array 10855718 - -
          Affymetrix Rat Gene 1.0 ST Array 10855716 - -
          Affymetrix Rat Gene 1.0 ST Array 10855725 - -
          Affymetrix Rat Gene 1.0 ST Array 10855726 - -
          Affymetrix Rat Gene 1.0 ST Array 10855715 - -
          Affymetrix Rat Gene 1.0 ST Array 10855720 - -
          Affymetrix Rat Gene 1.1 ST Array 10855707 - -
          Rat Genome 230 2.0 Array 1369494_a_at - -
          Rat Genome 230 2.0 Array 1369494_PM_a_at - -
          Rat Genome 230 2.0 Array 1370730_PM_a_at - -
          Rat Genome 230 2.0 Array 1370730_a_at - -
          Affymetrix HT Rat Focus Array 1369494_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369494_a_at - -
          Affymetrix HT Rat Focus Array 1370730_a_at - -
          Affymetrix HT Rat Focus Array 1370730_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400006972.rn.1 TC0400000975.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006971.rn.1 TC0400000975.rn.1 RTA TC
          Rat Expression Set 230 1370730_a_at - -
          Rat Expression Set 230 1370730_PM_a_at - -
          Rat Expression Set 230 1369494_a_at - -
          Rat Expression Set 230 1369494_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855707 - -
          Rat Genome U34 Set L07380_g_at - -
          Rat Genome U34 Set L07380_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21005802 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368059 - -
          RatRef-12 Expression BeadChip ILMN_48338 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline