Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563585_m1
          See other CNTN2 GE Assays ›
          Gene Symbol:
          Cntn2
          Gene Name:
          contactin 2 (axonal)
          Gene Aliases:
          TAG-1, TAG-564, TAG1, Tax
          Chromosome Location:
          Chr.13: 49280913 - 49314047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563585_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cntn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012884.1 NP_037016.1 16 - 17 2349 57
          XM_006249746.2 XP_006249808.1 17 - 18 2563 57
          XM_008769427.1 XP_008767649.1 16 - 17 2449 57
          GenBank mRNA M31725.1 - 16 - 17 2349 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001404.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cntn2

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          neuron migration
          cell-matrix adhesion
          axonogenesis
          axon guidance
          axonal fasciculation
          learning
          adult walking behavior
          regulation of cell morphogenesis involved in differentiation
          positive regulation of protein processing
          cerebral cortex GABAergic interneuron migration
          central nervous system myelination
          regulation of axon diameter
          neuron projection development
          receptor internalization
          cellular protein localization
          clustering of voltage-gated potassium channels
          negative regulation of neuron differentiation
          regulation of neuronal synaptic plasticity
          regulation of astrocyte differentiation
          positive regulation of adenosine receptor signaling pathway
          protein localization to juxtaparanode region of axon
          establishment of protein localization to juxtaparanode region of axon
          presynaptic membrane organization
          glycoprotein binding
          carbohydrate binding
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P434271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132218 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5153 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434271 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P434271 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P434271 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P434271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767622 - -
          Affymetrix Rat Gene 1.0 ST Array 10767623 - -
          Affymetrix Rat Gene 1.0 ST Array 10767626 - -
          Affymetrix Rat Gene 1.0 ST Array 10767611 - -
          Affymetrix Rat Gene 1.0 ST Array 10767620 - -
          Affymetrix Rat Gene 1.0 ST Array 10767628 - -
          Affymetrix Rat Gene 1.0 ST Array 10767607 - -
          Affymetrix Rat Gene 1.0 ST Array 10767621 - -
          Affymetrix Rat Gene 1.0 ST Array 10767624 - -
          Affymetrix Rat Gene 1.0 ST Array 10767608 - -
          Affymetrix Rat Gene 1.0 ST Array 10767619 - -
          Affymetrix Rat Gene 1.0 ST Array 10767610 - -
          Affymetrix Rat Gene 1.0 ST Array 10767627 - -
          Affymetrix Rat Gene 1.0 ST Array 10767616 - -
          Affymetrix Rat Gene 1.0 ST Array 10767617 - -
          Affymetrix Rat Gene 1.0 ST Array 10767618 - -
          Affymetrix Rat Gene 1.0 ST Array 10767606 - -
          Affymetrix Rat Gene 1.0 ST Array 10767625 - -
          Affymetrix Rat Gene 1.0 ST Array 10767609 - -
          Affymetrix Rat Gene 1.0 ST Array 10767612 - -
          Affymetrix Rat Gene 1.0 ST Array 10767615 - -
          Affymetrix Rat Gene 1.0 ST Array 10767613 - -
          Affymetrix Rat Gene 1.0 ST Array 10767614 - -
          Affymetrix Rat Gene 1.1 ST Array 10767605 - -
          Rat Genome 230 2.0 Array 1387424_at - -
          Rat Genome 230 2.0 Array 1387424_PM_at - -
          Affymetrix HT Rat Focus Array 1387424_PM_at - -
          Affymetrix HT Rat Focus Array 1387424_at - -
          Affymetrix Rat Clariom S Assay PSR1300009309.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009310.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009320.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009327.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009331.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009337.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009313.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009317.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009319.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009324.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009325.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009326.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009328.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009332.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009333.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009322.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009329.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009312.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009315.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009316.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009334.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009311.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009318.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009336.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009314.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009323.rn.1 TC1300001404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300009330.rn.1 TC1300001404.rn.1 RTA TC
          Rat Expression Set 230 1387424_at - -
          Rat Expression Set 230 1387424_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767605 - -
          Rat Genome U34 Set M31725_at - -
          Rat Genome U34 Set M31725_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20884449 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367489 - -
          RatRef-12 Expression BeadChip ILMN_66367 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline