Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563754_m1
          See other FASLG GE Assays ›
          Gene Symbol:
          Faslg
          Gene Name:
          Fas ligand (TNF superfamily, member 6)
          Gene Aliases:
          Apt1Lg1, CD95-L, Fasl, Tnfsf6
          Chromosome Location:
          Chr.13: 79696813 - 79716821 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563754_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Faslg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012908.1 NP_037040.1 3 - 4 517 101
          XM_006250148.2 XP_006250210.1 6 - 7 1156 101
          XM_008769607.1 XP_008767829.1 7 - 8 1391 101
          GenBank mRNA U03470.1 - 3 - 4 517 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001758.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Faslg

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory cell apoptotic process
          immune response
          positive regulation of cell proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of angiogenesis
          cellular chloride ion homeostasis
          response to lipopolysaccharide
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of neuron apoptotic process
          positive regulation of epidermal growth factor receptor signaling pathway
          retinal cell programmed cell death
          endosomal lumen acidification
          T cell apoptotic process
          necroptotic process
          response to growth factor
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          necroptotic signaling pathway
          positive regulation of endothelial cell apoptotic process
          death receptor binding
          cytokine activity
          tumor necrosis factor receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P409820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P409820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P409820 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68128 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769234 - -
          Affymetrix Rat Gene 1.0 ST Array 10769233 - -
          Affymetrix Rat Gene 1.0 ST Array 10769232 - -
          Affymetrix Rat Gene 1.0 ST Array 10769235 - -
          Affymetrix Rat Gene 1.1 ST Array 10769231 - -
          Rat Genome 230 2.0 Array 1387587_PM_at - -
          Rat Genome 230 2.0 Array 1387587_at - -
          Affymetrix HT Rat Focus Array 1387587_PM_at - -
          Affymetrix HT Rat Focus Array 1387587_at - -
          Affymetrix Rat Clariom S Assay PSR1300012309.rn.1 TC1300001758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012307.rn.1 TC1300001758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012306.rn.1 TC1300001758.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300012308.rn.1 TC1300001758.rn.1 RTA TC
          Rat Expression Set 230 1387587_PM_at - -
          Rat Expression Set 230 1387587_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769231 - -
          Rat Genome U34 Set U03470_at - -
          Rat Genome U34 Set U03470_at - -
          Rat Genome U34 Set U03470_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22136990 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372914 - -
          RatRef-12 Expression BeadChip ILMN_68070 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline