Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563784_m1
          See other ATF3 GE Assays ›
          Gene Symbol:
          Atf3
          Gene Name:
          activating transcription factor 3
          Gene Aliases:
          LRF-1, LRFI
          Chromosome Location:
          Chr.13: 109817892 - 109849632 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563784_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012912.2 NP_037044.1 3 - 4 510 88
          GenBank mRNA BC078903.1 - 3 - 4 510 7100767 88
          M63282.1 - 3 - 4 515 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002212.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf3

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell fate determination
          gluconeogenesis
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          skeletal muscle cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          identical protein binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152115 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38221 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11616 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11616 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11616 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73786 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770713 - -
          Affymetrix Rat Gene 1.0 ST Array 10770712 - -
          Affymetrix Rat Gene 1.0 ST Array 10770711 - -
          Affymetrix Rat Gene 1.1 ST Array 10770710 - -
          Rat Genome 230 2.0 Array 1369268_PM_at - -
          Rat Genome 230 2.0 Array 1369268_at - -
          Affymetrix HT Rat Focus Array 1369268_at - -
          Affymetrix HT Rat Focus Array 1369268_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300015757.rn.1 TC1300002212.rn.1 RTA TC
          Rat Expression Set 230 1369268_at - -
          Rat Expression Set 230 1369268_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770710 - -
          Rat Genome U34 Set M63282_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21030397 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363083 - -
          RatRef-12 Expression BeadChip ILMN_55728 - -

          Back To Top
           

          Citations (2)

          SOCS3-mediated blockade of JAK/STAT3 signaling pathway reveals its major contribution to spinal cord neuroinflammation and mechanical allodynia after peripheral nerve injury.
          Authors
          Dominguez E, Mauborgne A, Mallet J, Desclaux M, Pohl M
          Journal
          J Neurosci - Page No 5754 (Wed Apr 21 00:00:00 EDT 2010)
          Volume (Year)
          30 (2010)
          Product Usage
          -
          Differential implication of proinflammatory cytokine interleukin-6 in the development of cephalic versus extracephalic neuropathic pain in rats.
          Authors
          Latremoliere A, Mauborgne A, Masson J, Bourgoin S, Kayser V, Hamon M, Pohl M
          Journal
          J Neurosci - Page No 8489 (Wed Aug 20 00:00:00 EDT 2008)
          Volume (Year)
          28 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0