Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563935_m1
          See other CDH6 GE Assays ›
          Gene Symbol:
          Cdh6
          Gene Name:
          cadherin 6, type 2, K-cadherin
          Gene Aliases:
          KCAD
          Chromosome Location:
          Chr.2: 63100723 - 63246618 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563935_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012927.1 NP_037059.1 2 - 3 414 105
          GenBank mRNA D25290.1 - 2 - 3 414 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003106.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdh6

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          female gonad development
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P141919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26854 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77890 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46595 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113800 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P141919 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P141919 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P141919 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P141919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22981 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52270 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99804 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65626 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822048 - -
          Affymetrix Rat Gene 1.0 ST Array 10822046 - -
          Affymetrix Rat Gene 1.0 ST Array 10822052 - -
          Affymetrix Rat Gene 1.0 ST Array 10822047 - -
          Affymetrix Rat Gene 1.0 ST Array 10822053 - -
          Affymetrix Rat Gene 1.0 ST Array 10822051 - -
          Affymetrix Rat Gene 1.0 ST Array 10822054 - -
          Affymetrix Rat Gene 1.0 ST Array 10822049 - -
          Affymetrix Rat Gene 1.0 ST Array 10822044 - -
          Affymetrix Rat Gene 1.0 ST Array 10822055 - -
          Affymetrix Rat Gene 1.0 ST Array 10822050 - -
          Affymetrix Rat Gene 1.1 ST Array 10822043 - -
          Rat Genome 230 2.0 Array 1370657_at - -
          Rat Genome 230 2.0 Array 1370657_PM_at - -
          Affymetrix HT Rat Focus Array 1370657_at - -
          Affymetrix HT Rat Focus Array 1370657_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200021745.rn.1 TC0200003106.rn.1 RTA TC
          Rat Expression Set 230 1370657_at - -
          Rat Expression Set 230 1370657_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822043 - -
          Rat Genome U34 Set D25290_at - -
          Rat Genome U34 Set D25290_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21918500 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline