Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00563995_m1
          See other CPT2 GE Assays ›
          Gene Symbol:
          Cpt2
          Gene Name:
          carnitine palmitoyltransferase 2
          Gene Aliases:
          CPTII
          Chromosome Location:
          Chr.5: 127505646 - 127523016 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00563995_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012930.1 NP_037062.1 1 - 2 214 89
          GenBank mRNA J05470.1 - 1 - 2 214 89
          U88295.1 - 1 - 2 152 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003315.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cpt2

          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid beta-oxidation
          metabolic process
          long-chain fatty acid transport
          carnitine O-palmitoyltransferase activity
          transferase activity, transferring acyl groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P601920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57499 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P601920 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P601920 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P601920 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P601920 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700597939 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2699 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49377 - -
          Agilent Rat cDNA Microarray (G4105A) 700597939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10878567 - -
          Affymetrix Rat Gene 1.0 ST Array 10878569 - -
          Affymetrix Rat Gene 1.0 ST Array 10878565 - -
          Affymetrix Rat Gene 1.0 ST Array 10878568 - -
          Affymetrix Rat Gene 1.0 ST Array 10878566 - -
          Affymetrix Rat Gene 1.0 ST Array 10878571 - -
          Affymetrix Rat Gene 1.0 ST Array 10878570 - -
          Affymetrix Rat Gene 1.1 ST Array 10878564 - -
          Rat Genome 230 2.0 Array 1386927_PM_at - -
          Rat Genome 230 2.0 Array 1386927_at - -
          Affymetrix HT Rat Focus Array 1386927_PM_at - -
          Affymetrix HT Rat Focus Array 1386927_at - -
          Affymetrix Rat Clariom S Assay PSR0500024089.rn.1 TC0500003315.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500024088.rn.1 TC0500003315.rn.1 RTA TC
          Rat Expression Set 230 1386927_PM_at - -
          Rat Expression Set 230 1386927_at - -
          Affymetrix Rat Gene 1.0 ST Array 10878564 - -
          Rat Genome U34 Set J05470_at - -
          Rat Genome U34 Set rc_AI058968_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21331455 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354686 - -
          RatRef-12 Expression BeadChip ILMN_69382 - -

          Back To Top
           

          Citations (1)

          Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
          Authors
          Gudbrandsen OA, Rost TH, Berge RK
          Journal
          J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          47 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline