Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564006_m1
          See other CRMP1 GE Assays ›
          Gene Symbol:
          Crmp1
          Gene Name:
          collapsin response mediator protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 78266931 - 78312778 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564006_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crmp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012932.2 NP_037064.1 12 - 13 1670 63
          XM_006251084.2 XP_006251146.1 12 - 13 1904 63
          GenBank mRNA U52095.1 - 12 - 13 1464 63
          U52102.1 - 12 - 13 1638 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002038.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crmp1

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          pyrimidine nucleobase catabolic process
          neuron development
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P336226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31607 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P336226 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P336226 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P336226 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P336226 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700182493 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103441 - -
          Agilent Rat cDNA Microarray (G4105A) 700182493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777396 - -
          Affymetrix Rat Gene 1.0 ST Array 10777391 - -
          Affymetrix Rat Gene 1.0 ST Array 10777387 - -
          Affymetrix Rat Gene 1.0 ST Array 10777384 - -
          Affymetrix Rat Gene 1.0 ST Array 10777392 - -
          Affymetrix Rat Gene 1.0 ST Array 10777380 - -
          Affymetrix Rat Gene 1.0 ST Array 10777389 - -
          Affymetrix Rat Gene 1.0 ST Array 10777385 - -
          Affymetrix Rat Gene 1.0 ST Array 10777390 - -
          Affymetrix Rat Gene 1.0 ST Array 10777381 - -
          Affymetrix Rat Gene 1.0 ST Array 10777388 - -
          Affymetrix Rat Gene 1.0 ST Array 10777379 - -
          Affymetrix Rat Gene 1.0 ST Array 10777393 - -
          Affymetrix Rat Gene 1.0 ST Array 10777394 - -
          Affymetrix Rat Gene 1.0 ST Array 10777383 - -
          Affymetrix Rat Gene 1.0 ST Array 10777382 - -
          Affymetrix Rat Gene 1.0 ST Array 10777395 - -
          Affymetrix Rat Gene 1.1 ST Array 10777378 - -
          Rat Genome 230 2.0 Array 1387899_PM_at - -
          Rat Genome 230 2.0 Array 1387899_at - -
          Affymetrix HT Rat Focus Array 1387899_PM_at - -
          Affymetrix HT Rat Focus Array 1387899_at - -
          Affymetrix Rat Clariom S Assay PSR1400014447.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014452.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014458.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014450.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014460.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014465.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014451.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014448.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014466.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014459.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014462.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014457.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014449.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014453.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014456.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014463.rn.1 TC1400002038.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014464.rn.1 TC1400002038.rn.1 RTA TC
          Rat Expression Set 230 1387899_PM_at - -
          Rat Expression Set 230 1387899_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777378 - -
          Rat Genome U34 Set U52102_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22098774 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374446 - -
          RatRef-12 Expression BeadChip ILMN_69016 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline