Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564052_m1
          See other CTSH GE Assays ›
          Gene Symbol:
          Ctsh
          Gene Name:
          cathepsin H
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 97439153 - 97458293 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564052_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012939.1 NP_037071.1 7 - 8 546 83
          XM_008766440.1 XP_008764662.1 7 - 8 597 83
          GenBank mRNA BC085352.1 - 7 - 8 561 7115929 83
          FQ210730.1 - 7 - 8 556 83
          FQ213183.1 - 7 - 8 556 83
          FQ219177.1 - 7 - 8 83
          FQ219285.1 - 7 - 8 556 83
          FQ219548.1 - 7 - 8 556 83
          FQ219611.1 - 7 - 8 574 83
          FQ220588.1 - 4 - 5 325 83
          FQ223707.1 - 7 - 8 552 83
          FQ228686.1 - 7 - 8 616 83
          FQ228906.1 - 7 - 8 563 83
          FQ228938.1 - 7 - 8 553 83
          M38135.1 - 7 - 8 540 83
          Y00708.1 - 7 - 8 546 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001277.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsh

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          T cell mediated cytotoxicity
          adaptive immune response
          immune response-regulating signaling pathway
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          spermatogenesis
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          neuropeptide catabolic process
          bradykinin catabolic process
          positive regulation of peptidase activity
          positive regulation of cell migration
          zymogen activation
          protein destabilization
          response to retinoic acid
          membrane protein proteolysis
          negative regulation of apoptotic process
          surfactant homeostasis
          positive regulation of angiogenesis
          dichotomous subdivision of terminal units involved in lung branching
          ERK1 and ERK2 cascade
          cellular response to thyroid hormone stimulus
          positive regulation of apoptotic signaling pathway
          endopeptidase activity
          aminopeptidase activity
          cysteine-type endopeptidase activity
          serine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          peptidase activator activity involved in apoptotic process
          HLA-A specific activating MHC class I receptor activity
          kininogen binding
          protein complex binding
          protein self-association
          thyroid hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P552849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P552849 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P552849 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P552849 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P552849 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700783730 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702147645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97105 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11018 - -
          Agilent Rat cDNA Microarray (G4105A) 700783730 - -
          Agilent Rat cDNA Microarray (G4105A) 702147645 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912172 - -
          Affymetrix Rat Gene 1.0 ST Array 10912164 - -
          Affymetrix Rat Gene 1.0 ST Array 10912166 - -
          Affymetrix Rat Gene 1.0 ST Array 10912170 - -
          Affymetrix Rat Gene 1.0 ST Array 10912165 - -
          Affymetrix Rat Gene 1.0 ST Array 10912167 - -
          Affymetrix Rat Gene 1.0 ST Array 10912171 - -
          Affymetrix Rat Gene 1.0 ST Array 10912168 - -
          Affymetrix Rat Gene 1.0 ST Array 10912173 - -
          Affymetrix Rat Gene 1.0 ST Array 10912162 - -
          Affymetrix Rat Gene 1.0 ST Array 10912169 - -
          Affymetrix Rat Gene 1.0 ST Array 10912163 - -
          Affymetrix Rat Gene 1.1 ST Array 10912161 - -
          Rat Genome 230 2.0 Array 1386899_at - -
          Rat Genome 230 2.0 Array 1386899_PM_at - -
          Affymetrix HT Rat Focus Array 1386899_PM_at - -
          Affymetrix HT Rat Focus Array 1386899_at - -
          Affymetrix Rat Clariom S Assay PSR0800009316.rn.1 TC0800001277.rn.1 RTA TC
          Rat Expression Set 230 1386899_PM_at - -
          Rat Expression Set 230 1386899_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912161 - -
          Rat Genome U34 Set M38135_at - -
          Rat Genome U34 Set rc_AI009415_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21861231 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367864 - -
          RatRef-12 Expression BeadChip ILMN_59666 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline