Search
Search

Sparcl1| Gene Symbol: Sparcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012946.1 | NP_037078.1 | 2 - 3 | 404 | 78 | |
| GenBank mRNA | BC061755.1 | - | 2 - 3 | 405 | 5600008 | 78 |
| U27562.1 | - | 2 - 3 | 404 | 78 | ||
Set Membership: |
|