Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564115_m1
          See other FGF10 GE Assays ›
          Gene Symbol:
          Fgf10
          Gene Name:
          fibroblast growth factor 10
          Gene Aliases:
          FGF-10
          Chromosome Location:
          Chr.2: 51673480 - 51747533 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564115_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012951.1 NP_037083.1 2 - 3 447 148
          GenBank mRNA D79215.1 - 2 - 3 447 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000461.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf10

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          activation of MAPK activity
          angiogenesis
          metanephros development
          organ induction
          mesonephros development
          blood vessel remodeling
          metanephros morphogenesis
          chemotaxis
          cell-cell signaling
          determination of left/right symmetry
          salivary gland development
          salivary gland morphogenesis
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          epidermis development
          regulation of smoothened signaling pathway
          embryonic pattern specification
          organ morphogenesis
          regulation of gene expression
          epithelial cell migration
          positive regulation of epithelial cell migration
          positive regulation of keratinocyte proliferation
          response to organic cyclic compound
          pituitary gland development
          cell differentiation
          positive regulation of Wnt signaling pathway
          lung development
          embryonic genitalia morphogenesis
          epithelial cell differentiation
          thyroid gland development
          otic vesicle formation
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          pancreas development
          hair follicle morphogenesis
          embryonic camera-type eye development
          actin cytoskeleton reorganization
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          response to estradiol
          response to lipopolysaccharide
          positive regulation of ATPase activity
          lacrimal gland development
          regulation of activin receptor signaling pathway
          protein localization to cell surface
          somatic stem cell population maintenance
          limb morphogenesis
          organ growth
          wound healing
          tissue regeneration
          inner ear morphogenesis
          odontogenesis of dentin-containing tooth
          muscle cell fate commitment
          positive regulation of MAPK cascade
          keratinocyte proliferation
          negative regulation of cell differentiation
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of Notch signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of Ras protein signal transduction
          regulation of saliva secretion
          positive regulation of fibroblast proliferation
          lung alveolus development
          blood vessel morphogenesis
          spleen development
          thymus development
          embryonic digestive tract morphogenesis
          digestive tract development
          embryonic digestive tract development
          organ formation
          epidermis morphogenesis
          semicircular canal morphogenesis
          branching morphogenesis of an epithelial tube
          female genitalia morphogenesis
          male genitalia morphogenesis
          positive regulation of lymphocyte proliferation
          epithelial cell proliferation
          urothelial cell proliferation
          positive regulation of urothelial cell proliferation
          regulation of epithelial cell proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          white fat cell differentiation
          positive chemotaxis
          induction of positive chemotaxis
          smooth muscle cell differentiation
          positive regulation of keratinocyte migration
          radial glial cell differentiation
          limb development
          limb bud formation
          lung morphogenesis
          lung epithelium development
          lung saccule development
          bronchiole morphogenesis
          epithelial tube branching involved in lung morphogenesis
          branching involved in salivary gland morphogenesis
          bud outgrowth involved in lung branching
          bud elongation involved in lung branching
          mesenchymal-epithelial cell signaling involved in lung development
          Type II pneumocyte differentiation
          prostatic bud formation
          respiratory system development
          mammary gland specification
          fibroblast growth factor receptor signaling pathway involved in mammary gland specification
          mammary gland bud formation
          submandibular salivary gland formation
          epithelial cell proliferation involved in salivary gland morphogenesis
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          branch elongation involved in salivary gland morphogenesis
          semicircular canal fusion
          mesenchymal cell differentiation involved in lung development
          secretion by lung epithelial cell involved in lung growth
          lung proximal/distal axis specification
          tear secretion
          positive regulation of white fat cell proliferation
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          Harderian gland development
          negative regulation of cell cycle arrest
          positive regulation of hair follicle cell proliferation
          positive regulation of canonical Wnt signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          growth factor activity
          heparin binding
          chemoattractant activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101309 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11630 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95640 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813174 - -
          Affymetrix Rat Gene 1.0 ST Array 10813173 - -
          Affymetrix Rat Gene 1.0 ST Array 10813175 - -
          Affymetrix Rat Gene 1.1 ST Array 10813172 - -
          Rat Genome 230 2.0 Array 1369604_at - -
          Rat Genome 230 2.0 Array 1392432_at - -
          Rat Genome 230 2.0 Array 1392432_PM_at - -
          Rat Genome 230 2.0 Array 1369604_PM_at - -
          Affymetrix HT Rat Focus Array 1369604_PM_at - -
          Affymetrix HT Rat Focus Array 1369604_at - -
          Affymetrix Rat Clariom S Assay PSR0200002628.rn.1 TC0200000461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002629.rn.1 TC0200000461.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002630.rn.1 TC0200000461.rn.1 RTA TC
          Rat Expression Set 230 1369604_PM_at - -
          Rat Expression Set 230 1369604_at - -
          Rat Expression Set 230 1392432_PM_at - -
          Rat Expression Set 230 1392432_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813172 - -
          Rat Genome U34 Set D79215_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22054844 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370793 - -
          RatRef-12 Expression BeadChip ILMN_53095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline