Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564119_m1
          See other FOSL1 GE Assays ›
          Gene Symbol:
          Fosl1
          Gene Name:
          fos-like antigen 1
          Gene Aliases:
          Fra-1, fra-1
          Chromosome Location:
          Chr.1: 220826560 - 220835066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564119_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fosl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012953.1 NP_037085.1 2 - 3 523 94
          GenBank mRNA BC097342.1 - 2 - 3 502 7459558 94
          M19651.1 - 2 - 3 523 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003054.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fosl1

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          vitellogenesis
          female pregnancy
          learning
          negative regulation of cell proliferation
          response to mechanical stimulus
          response to gravity
          response to organic cyclic compound
          cellular response to extracellular stimulus
          response to progesterone
          response to cytokine
          response to drug
          response to hydrogen peroxide
          positive regulation of apoptotic process
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          response to corticosterone
          response to cAMP
          placenta blood vessel development
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596131 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100321 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123118 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P693821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100251 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88705 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713048 - -
          Affymetrix Rat Gene 1.0 ST Array 10713046 - -
          Affymetrix Rat Gene 1.0 ST Array 10713049 - -
          Affymetrix Rat Gene 1.0 ST Array 10713047 - -
          Affymetrix Rat Gene 1.1 ST Array 10713045 - -
          Rat Genome 230 2.0 Array 1368489_PM_at - -
          Rat Genome 230 2.0 Array 1368489_at - -
          Affymetrix HT Rat Focus Array 1368489_at - -
          Affymetrix HT Rat Focus Array 1368489_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100024736.rn.1 TC0100003054.rn.1 RTA TC
          Rat Expression Set 230 1368489_at - -
          Rat Expression Set 230 1368489_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713045 - -
          Rat Genome U34 Set M19651_at - -
          Rat Genome U34 Set M19651_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21305806 - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_54965 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline