Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564149_m1
          See other GABRB2 GE Assays ›
          Gene Symbol:
          Gabrb2
          Gene Name:
          gamma-aminobutyric acid (GABA) A receptor, beta 2
          Gene Aliases:
          GARB2
          Chromosome Location:
          Chr.10: 27973692 - 28193072 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564149_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabrb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012957.2 NP_037089.1 4 - 5 454 64
          XM_006246127.2 XP_006246189.1 5 - 6 915 64
          XM_008767635.1 XP_008765857.1 5 - 6 919 64
          GenBank mRNA AY574251.1 - 4 - 5 454 64
          X15467.1 - 4 - 5 530 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000407.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gabrb2

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          gamma-aminobutyric acid signaling pathway
          sensory perception of sound
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          neuron development
          synaptic transmission, GABAergic
          inner ear receptor cell development
          innervation
          cellular response to histamine
          cochlea development
          negative regulation of neuron death
          chloride transmembrane transport
          GABA-A receptor activity
          inhibitory extracellular ligand-gated ion channel activity
          chloride channel activity
          protein binding
          GABA receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P099059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60768 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132478 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111876 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31869 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58871 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P099059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P099059 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P641587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18733 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76504 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58080 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732832 - -
          Affymetrix Rat Gene 1.0 ST Array 10732833 - -
          Affymetrix Rat Gene 1.0 ST Array 10732826 - -
          Affymetrix Rat Gene 1.0 ST Array 10732829 - -
          Affymetrix Rat Gene 1.0 ST Array 10732831 - -
          Affymetrix Rat Gene 1.0 ST Array 10732827 - -
          Affymetrix Rat Gene 1.0 ST Array 10732830 - -
          Affymetrix Rat Gene 1.0 ST Array 10732835 - -
          Affymetrix Rat Gene 1.0 ST Array 10732828 - -
          Affymetrix Rat Gene 1.1 ST Array 10732824 - -
          Rat Genome 230 2.0 Array 1369818_PM_at - -
          Rat Genome 230 2.0 Array 1369818_at - -
          Affymetrix HT Rat Focus Array 1369818_PM_at - -
          Affymetrix HT Rat Focus Array 1369818_at - -
          Affymetrix Rat Clariom S Assay PSR1000003518.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003522.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003520.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003521.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003515.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003516.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003517.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003519.rn.1 TC1000000407.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000003523.rn.1 TC1000000407.rn.1 RTA TC
          Rat Expression Set 230 1369818_at - -
          Rat Expression Set 230 1369818_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732824 - -
          Rat Genome U34 Set X15467cds_at - -
          Rat Genome U34 Set X15467cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21231706 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372713 - -
          RatRef-12 Expression BeadChip ILMN_50751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline