Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564204_m1
          See other HMMR GE Assays ›
          Gene Symbol:
          Hmmr
          Gene Name:
          hyaluronan mediated motility receptor (RHAMM)
          Gene Aliases:
          RHAMM
          Chromosome Location:
          Chr.10: 25861490 - 25890639 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564204_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmmr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012964.2 NP_037096.2 17 - 18 2147 71
          GenBank mRNA AF133037.1 - 17 - 18 2147 71
          AF336825.1 - 17 - 18 2147 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002592.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmmr

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          cell-cell signaling
          hyaluronic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P351118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P351118 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P351118 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P351118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P351118 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P292314 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742132 - -
          Affymetrix Rat Gene 1.0 ST Array 10742144 - -
          Affymetrix Rat Gene 1.0 ST Array 10742134 - -
          Affymetrix Rat Gene 1.0 ST Array 10742133 - -
          Affymetrix Rat Gene 1.0 ST Array 10742142 - -
          Affymetrix Rat Gene 1.0 ST Array 10742131 - -
          Affymetrix Rat Gene 1.0 ST Array 10742137 - -
          Affymetrix Rat Gene 1.0 ST Array 10742143 - -
          Affymetrix Rat Gene 1.0 ST Array 10742138 - -
          Affymetrix Rat Gene 1.0 ST Array 10742135 - -
          Affymetrix Rat Gene 1.0 ST Array 10742139 - -
          Affymetrix Rat Gene 1.0 ST Array 10742130 - -
          Affymetrix Rat Gene 1.0 ST Array 10742140 - -
          Affymetrix Rat Gene 1.0 ST Array 10742136 - -
          Affymetrix Rat Gene 1.0 ST Array 10742129 - -
          Affymetrix Rat Gene 1.1 ST Array 10742128 - -
          Rat Genome 230 2.0 Array 1370461_at - -
          Rat Genome 230 2.0 Array 1370462_PM_at - -
          Rat Genome 230 2.0 Array 1370462_at - -
          Rat Genome 230 2.0 Array 1370461_PM_at - -
          Affymetrix HT Rat Focus Array 1370462_at - -
          Affymetrix HT Rat Focus Array 1370461_PM_at - -
          Affymetrix HT Rat Focus Array 1370461_at - -
          Affymetrix HT Rat Focus Array 1370462_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000023170.rn.1 TC1000002592.rn.1 RTA TC
          Rat Expression Set 230 1370462_at - -
          Rat Expression Set 230 1370462_PM_at - -
          Rat Expression Set 230 1370461_PM_at - -
          Rat Expression Set 230 1370461_at - -
          Affymetrix Rat Gene 1.0 ST Array 10742128 - -
          Rat Genome U34 Set U87983_at - -
          Rat Genome U34 Set rc_AI171185_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21388641 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350866 - -
          RatRef-12 Expression BeadChip ILMN_68349 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline