Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564309_m1
          See other LHCGR GE Assays ›
          Gene Symbol:
          Lhcgr
          Gene Name:
          luteinizing hormone/choriogonadotropin receptor
          Gene Aliases:
          LH/CG-R, LSH-R, LSHR, Lhr
          Chromosome Location:
          Chr.6: 12493182 - 12554482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564309_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lhcgr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012978.1 NP_037110.1 9 - 10 1003 77
          GenBank mRNA M26199.1 - 9 - 10 1003 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001901.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lhcgr

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          protein targeting to lysosome
          G-protein coupled receptor signaling pathway
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          phospholipase C-activating G-protein coupled receptor signaling pathway
          spermatogenesis
          central nervous system development
          male gonad development
          female gonad development
          hormone-mediated signaling pathway
          positive regulation of calcium ion transport into cytosol
          ovulation cycle process
          positive regulation of inositol trisphosphate biosynthetic process
          response to drug
          luteinizing hormone signaling pathway
          positive regulation of cAMP-mediated signaling
          development of secondary male sexual characteristics
          positive regulation of hormone biosynthetic process
          arachidonic acid secretion
          regulation of steroid biosynthetic process
          positive regulation of calcium-mediated signaling
          cognition
          positive regulation of release of sequestered calcium ion into cytosol
          uterus development
          cellular response to gonadotropin stimulus
          seminiferous tubule development
          regulation of steroid hormone biosynthetic process
          luteinizing hormone receptor activity
          G-protein coupled peptide receptor activity
          peptide hormone binding
          choriogonadotropin hormone receptor activity
          choriogonadotropin hormone binding
          identical protein binding
          protein homodimerization activity
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P130286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1648 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130286 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P130286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130286 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82539 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90867 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10888346 - -
          Affymetrix Rat Gene 1.0 ST Array 10888348 - -
          Affymetrix Rat Gene 1.0 ST Array 10888351 - -
          Affymetrix Rat Gene 1.0 ST Array 10888350 - -
          Affymetrix Rat Gene 1.0 ST Array 10888352 - -
          Affymetrix Rat Gene 1.0 ST Array 10888345 - -
          Affymetrix Rat Gene 1.0 ST Array 10888353 - -
          Affymetrix Rat Gene 1.0 ST Array 10888344 - -
          Affymetrix Rat Gene 1.0 ST Array 10888347 - -
          Affymetrix Rat Gene 1.0 ST Array 10888349 - -
          Affymetrix Rat Gene 1.1 ST Array 10888343 - -
          Rat Genome 230 2.0 Array 1387423_at - -
          Rat Genome 230 2.0 Array 1369033_at - -
          Rat Genome 230 2.0 Array 1387423_PM_at - -
          Rat Genome 230 2.0 Array 1369033_PM_at - -
          Affymetrix HT Rat Focus Array 1369033_PM_at - -
          Affymetrix HT Rat Focus Array 1387423_PM_at - -
          Affymetrix HT Rat Focus Array 1387423_at - -
          Affymetrix HT Rat Focus Array 1369033_at - -
          Affymetrix Rat Clariom S Assay PSR0600013449.rn.1 TC0600001901.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600013450.rn.1 TC0600001901.rn.1 RTA TC
          Rat Expression Set 230 1387423_at - -
          Rat Expression Set 230 1369033_PM_at - -
          Rat Expression Set 230 1387423_PM_at - -
          Rat Expression Set 230 1369033_at - -
          Affymetrix Rat Gene 1.0 ST Array 10888343 - -
          Rat Genome U34 Set Z33403_at - -
          Rat Genome U34 Set M61211cds_s_at - -
          Rat Genome U34 Set M63926mRNA_s_at - -
          Rat Genome U34 Set rc_AI236945_s_at - -
          Rat Genome U34 Set M26199_s_at - -
          Rat Genome U34 Set S40803cds#3_s_at - -
          Rat Genome U34 Set rc_AI177137_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21678174 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371979 - -
          RatRef-12 Expression BeadChip ILMN_61765 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline