Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564562_m1
          See other POU1F1 GE Assays ›
          Gene Symbol:
          Pou1f1
          Gene Name:
          POU class 1 homeobox 1
          Gene Aliases:
          GHF1, GHF1A, PIT1Z, Pit1
          Chromosome Location:
          Chr.11: 2645659 - 2662581 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564562_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pou1f1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013008.3 NP_037140.2 2 - 3 333 75
          XM_006247974.1 XP_006248036.1 2 - 3 617 75
          GenBank mRNA AB622208.1 - 1 - 2 344 75
          BC061563.1 - 2 - 3 337 6918965 75
          BC061564.1 - 2 - 3 352 75
          L01506.1 - 2 - 3 301 75
          L01507.1 - 2 - 3 377 75
          M23253.1 - 2 - 3 247 75
          X12658.1 - 2 - 3 270 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000009.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pou1f1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell fate specification
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          determination of adult lifespan
          pituitary gland development
          adenohypophysis development
          B cell differentiation
          positive regulation of inositol trisphosphate biosynthetic process
          positive regulation of multicellular organism growth
          regulation of insulin-like growth factor receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          nuclear transport
          somatotropin secreting cell differentiation
          somatotropin secreting cell development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164627 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164627 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127971 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164627 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127971 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164627 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11655 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19594 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749830 - -
          Affymetrix Rat Gene 1.0 ST Array 10749838 - -
          Affymetrix Rat Gene 1.0 ST Array 10749827 - -
          Affymetrix Rat Gene 1.0 ST Array 10749833 - -
          Affymetrix Rat Gene 1.0 ST Array 10749836 - -
          Affymetrix Rat Gene 1.0 ST Array 10749828 - -
          Affymetrix Rat Gene 1.0 ST Array 10749835 - -
          Affymetrix Rat Gene 1.0 ST Array 10749837 - -
          Affymetrix Rat Gene 1.0 ST Array 10749829 - -
          Affymetrix Rat Gene 1.0 ST Array 10749834 - -
          Affymetrix Rat Gene 1.0 ST Array 10749832 - -
          Affymetrix Rat Gene 1.0 ST Array 10749831 - -
          Affymetrix Rat Gene 1.1 ST Array 10749826 - -
          Rat Genome 230 2.0 Array 1369761_PM_at - -
          Rat Genome 230 2.0 Array 1369762_at - -
          Rat Genome 230 2.0 Array 1369762_PM_at - -
          Rat Genome 230 2.0 Array 1369761_at - -
          Affymetrix HT Rat Focus Array 1369762_at - -
          Affymetrix HT Rat Focus Array 1369761_PM_at - -
          Affymetrix HT Rat Focus Array 1369762_PM_at - -
          Affymetrix HT Rat Focus Array 1369761_at - -
          Affymetrix Rat Clariom S Assay PSR1100000030.rn.1 TC1100000009.rn.1 RTA TC
          Rat Expression Set 230 1369762_PM_at - -
          Rat Expression Set 230 1369761_at - -
          Rat Expression Set 230 1369762_at - -
          Rat Expression Set 230 1369761_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749826 - -
          Rat Genome U34 Set L01507_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20708746 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356790 - -
          RatRef-12 Expression BeadChip ILMN_47778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline