Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564605_m1
          See other PTGDS GE Assays ›
          Gene Symbol:
          Ptgds
          Gene Name:
          prostaglandin D2 synthase (brain)
          Gene Aliases:
          PH2DISO
          Chromosome Location:
          Chr.3: 2686125 - 2689059 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564605_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptgds
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013015.2 NP_037147.1 1 - 2 148 57
          GenBank mRNA J04488.1 - 1 - 2 148 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002679.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptgds

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          transport
          regulation of circadian sleep/wake cycle, sleep
          response to glucocorticoid
          negative regulation of male germ cell proliferation
          prostaglandin-D synthase activity
          transporter activity
          retinoid binding
          fatty acid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30819 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15529 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15529 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15529 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843466 - -
          Affymetrix Rat Gene 1.0 ST Array 10843467 - -
          Affymetrix Rat Gene 1.0 ST Array 10843463 - -
          Affymetrix Rat Gene 1.0 ST Array 10843461 - -
          Affymetrix Rat Gene 1.0 ST Array 10843462 - -
          Affymetrix Rat Gene 1.0 ST Array 10843464 - -
          Affymetrix Rat Gene 1.0 ST Array 10843465 - -
          Affymetrix Rat Gene 1.1 ST Array 10843460 - -
          Rat Genome 230 2.0 Array 1367851_at - -
          Rat Genome 230 2.0 Array 1367851_PM_at - -
          Affymetrix HT Rat Focus Array 1367851_at - -
          Affymetrix HT Rat Focus Array 1367851_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019965.rn.1 TC0300002679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019962.rn.1 TC0300002679.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300019963.rn.1 TC0300002679.rn.1 RTA TC
          Rat Expression Set 230 1367851_at - -
          Rat Expression Set 230 1367851_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843460 - -
          Rat Genome U34 Set J04488_at - -
          Rat Genome U34 Set rc_AI144744_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372946 - -
          RatRef-12 Expression BeadChip ILMN_70000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline