Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564916_m1
          See other MAP3K12 GE Assays ›
          Gene Symbol:
          Map3k12
          Gene Name:
          mitogen activated protein kinase kinase kinase 12
          Gene Aliases:
          DLK, MUK, PK, Zpk
          Chromosome Location:
          Chr.7: 144102153 - 144120306 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564916_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map3k12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013055.1 NP_037187.1 4 - 5 891 72
          XM_006242381.2 XP_006242443.1 4 - 5 2272 72
          XM_006242382.2 XP_006242444.1 2 - 3 505 72
          XM_008765675.1 XP_008763897.1 1 - 2 1235 72
          GenBank mRNA D49785.1 - 4 - 5 891 72
          FQ211695.1 - 4 - 5 741 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003973.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Map3k12

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          protein phosphorylation
          JNK cascade
          histone phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          protein autophosphorylation
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          ATP binding
          protein kinase binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700032167.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032168.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032158.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032160.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032173.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032161.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032164.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032162.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032165.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032159.rn.1 TC0700003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700032163.rn.1 TC0700003973.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline