Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564925_m1
          See other F3 GE Assays ›
          Gene Symbol:
          F3
          Gene Name:
          coagulation factor III (thromboplastin, tissue factor)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 225310686 - 225322281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564925_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013057.2 NP_037189.2 5 - 6 851 58
          GenBank mRNA BC081846.1 - 5 - 6 851 7096602 58
          U07619.1 - 5 - 6 818 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002103.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene F3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          activation of plasma proteins involved in acute inflammatory response
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          aging
          blood coagulation
          response to temperature stimulus
          response to wounding
          response to mechanical stimulus
          positive regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of smooth muscle cell migration
          cytokine-mediated signaling pathway
          response to estradiol
          response to lipopolysaccharide
          response to fluid shear stress
          positive regulation of angiogenesis
          positive regulation of protein kinase B signaling
          response to low-density lipoprotein particle
          cellular response to hydrogen peroxide
          protease binding
          cytokine receptor activity
          phospholipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140551 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11683 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11683 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11683 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11683 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700591322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3471 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66099 - -
          Agilent Rat cDNA Microarray (G4105A) 700591322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818713 - -
          Affymetrix Rat Gene 1.0 ST Array 10818709 - -
          Affymetrix Rat Gene 1.0 ST Array 10818712 - -
          Affymetrix Rat Gene 1.0 ST Array 10818710 - -
          Affymetrix Rat Gene 1.0 ST Array 10818714 - -
          Affymetrix Rat Gene 1.0 ST Array 10818711 - -
          Affymetrix Rat Gene 1.1 ST Array 10818708 - -
          Rat Genome 230 2.0 Array 1369182_PM_at - -
          Rat Genome 230 2.0 Array 1369182_at - -
          Affymetrix HT Rat Focus Array 1369182_at - -
          Affymetrix HT Rat Focus Array 1369182_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200014474.rn.1 TC0200002103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014476.rn.1 TC0200002103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014473.rn.1 TC0200002103.rn.1 RTA TC
          Rat Expression Set 230 1369182_PM_at - -
          Rat Expression Set 230 1369182_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818708 - -
          Rat Genome U34 Set U07619_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21270816 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356793 - -
          RatRef-12 Expression BeadChip ILMN_60664 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline