Search
Search
| Gene Symbol: Id3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013058.2 | NP_037190.1 | 2 - 3 | 418 | 128 | |
| GenBank mRNA | BC064658.1 | - | 2 - 3 | 418 | 5623701 | 128 |
| D10864.1 | - | 2 - 3 | 395 | 128 | ||
| FQ212997.1 | - | 2 - 3 | 440 | 128 | ||
| FQ216267.1 | - | 2 - 3 | 440 | 128 | ||
| FQ218958.1 | - | 2 - 3 | 431 | 128 | ||
| FQ229332.1 | - | 2 - 3 | 440 | 128 | ||
Set Membership: |
|