Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564986_m1
          See other KDR GE Assays ›
          Gene Symbol:
          Kdr
          Gene Name:
          kinase insert domain receptor
          Gene Aliases:
          Vegfr-2
          Chromosome Location:
          Chr.14: 34727677 - 34787127 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564986_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013062.1 NP_037194.1 1 - 2 313 75
          GenBank mRNA BC087029.1 - 1 - 2 289 7124961 75
          U93306.1 - 1 - 2 313 75
          U93307.1 - 1 - 2 313 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000404.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kdr

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          ovarian follicle development
          vasculogenesis
          response to hypoxia
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          lymph vessel development
          cell migration involved in sprouting angiogenesis
          positive regulation of mesenchymal cell proliferation
          endocardium development
          endothelium development
          endochondral bone growth
          positive regulation of cytosolic calcium ion concentration
          aging
          positive regulation of cell proliferation
          regulation of cell shape
          male gonad development
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          hyaluronan metabolic process
          lung development
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          positive regulation of TOR signaling
          embryonic hemopoiesis
          calcium-mediated signaling using intracellular calcium source
          cellular response to vascular endothelial growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          vascular endothelial growth factor signaling pathway
          response to drug
          negative regulation of apoptotic process
          surfactant homeostasis
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          cell fate commitment
          endothelial cell differentiation
          regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of vasodilation
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          positive regulation of long-term neuronal synaptic plasticity
          lung alveolus development
          cell maturation
          branching morphogenesis of an epithelial tube
          neuron projection morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of neurogenesis
          positive regulation of calcium-mediated signaling
          positive regulation of positive chemotaxis
          positive regulation of nitric-oxide synthase biosynthetic process
          positive regulation of focal adhesion assembly
          calcium ion homeostasis
          positive regulation of ERK1 and ERK2 cascade
          response to oxygen levels
          regulation of hematopoietic progenitor cell differentiation
          negative regulation of endothelial cell apoptotic process
          positive regulation of vasculogenesis
          protein tyrosine kinase activity
          vascular endothelial growth factor-activated receptor activity
          integrin binding
          protein binding
          ATP binding
          growth factor binding
          vascular endothelial growth factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1400003162.rn.1 TC1400000404.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400003161.rn.1 TC1400000404.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21037155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline