Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565018_m1
          See other PARP1 GE Assays ›
          Gene Symbol:
          Parp1
          Gene Name:
          poly (ADP-ribose) polymerase 1
          Gene Aliases:
          ARTD1, Adprt, Parp-1
          Chromosome Location:
          Chr.13: 98857255 - 98889444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565018_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Parp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013063.2 NP_037195.1 2 - 3 345 70
          GenBank mRNA BC085765.1 - 2 - 3 345 7107130 70
          U94340.1 - 2 - 3 313 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000904.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Parp1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA metabolic process
          base-excision repair
          double-strand break repair
          transcription, DNA-templated
          protein ADP-ribosylation
          transforming growth factor beta receptor signaling pathway
          protein autoprocessing
          signal transduction involved in regulation of gene expression
          cellular response to insulin stimulus
          regulation of growth rate
          DNA damage response, detection of DNA damage
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of SMAD protein import into nucleus
          protein poly-ADP-ribosylation
          cellular response to superoxide
          positive regulation of transcription regulatory region DNA binding
          DNA binding
          NAD+ ADP-ribosyltransferase activity
          transcription factor binding
          zinc ion binding
          transferase activity, transferring pentosyl groups
          enzyme binding
          protein kinase binding
          identical protein binding
          poly(A) RNA binding
          protein N-terminus binding
          NAD binding
          R-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1033023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1836 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1033023 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1033023 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1033023 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11687 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1033023 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702021494 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103694 - -
          Agilent Rat cDNA Microarray (G4105A) 702021494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10766181 - -
          Affymetrix Rat Gene 1.0 ST Array 10766174 - -
          Affymetrix Rat Gene 1.0 ST Array 10766173 - -
          Affymetrix Rat Gene 1.0 ST Array 10766186 - -
          Affymetrix Rat Gene 1.0 ST Array 10766171 - -
          Affymetrix Rat Gene 1.0 ST Array 10766176 - -
          Affymetrix Rat Gene 1.0 ST Array 10766194 - -
          Affymetrix Rat Gene 1.0 ST Array 10766183 - -
          Affymetrix Rat Gene 1.0 ST Array 10766188 - -
          Affymetrix Rat Gene 1.0 ST Array 10766179 - -
          Affymetrix Rat Gene 1.0 ST Array 10766177 - -
          Affymetrix Rat Gene 1.0 ST Array 10766189 - -
          Affymetrix Rat Gene 1.0 ST Array 10766191 - -
          Affymetrix Rat Gene 1.0 ST Array 10766182 - -
          Affymetrix Rat Gene 1.0 ST Array 10766185 - -
          Affymetrix Rat Gene 1.0 ST Array 10766190 - -
          Affymetrix Rat Gene 1.0 ST Array 10766172 - -
          Affymetrix Rat Gene 1.0 ST Array 10766175 - -
          Affymetrix Rat Gene 1.0 ST Array 10766184 - -
          Affymetrix Rat Gene 1.0 ST Array 10766193 - -
          Affymetrix Rat Gene 1.0 ST Array 10766178 - -
          Affymetrix Rat Gene 1.0 ST Array 10766192 - -
          Affymetrix Rat Gene 1.0 ST Array 10766180 - -
          Affymetrix Rat Gene 1.0 ST Array 10766187 - -
          Affymetrix Rat Gene 1.1 ST Array 10766170 - -
          Rat Genome 230 2.0 Array 1369969_PM_at - -
          Rat Genome 230 2.0 Array 1369969_at - -
          Affymetrix HT Rat Focus Array 1369969_PM_at - -
          Affymetrix HT Rat Focus Array 1369969_at - -
          Affymetrix Rat Clariom S Assay PSR1300006352.rn.1 TC1300000904.rn.1 RTA TC
          Rat Expression Set 230 1369969_at - -
          Rat Expression Set 230 1369969_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10766170 - -
          Rat Genome U34 Set rc_AA964849_at - -
          Rat Genome U34 Set U94340_at - -
          Rat Genome U34 Set U94340_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21911201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360864 - -
          RatRef-12 Expression BeadChip ILMN_58369 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline