Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565155_m1
          See other HCRTR2 GE Assays ›
          Gene Symbol:
          Hcrtr2
          Gene Name:
          hypocretin (orexin) receptor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 83307684 - 83421972 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565155_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hcrtr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013074.1 NP_037206.1 6 - 7 1168 84
          XM_008766215.1 XP_008764437.1 6 - 7 1528 84
          GenBank mRNA AF041246.1 - 6 - 7 1168 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002952.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hcrtr2

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          feeding behavior
          circadian sleep/wake cycle process
          cellular response to hormone stimulus
          regulation of circadian sleep/wake cycle, sleep
          response to peptide
          G-protein coupled receptor activity
          neuropeptide receptor activity
          orexin receptor activity
          peptide hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P206844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71664 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109041 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91486 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153640 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50049 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206844 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206844 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P206844 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P206844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27966 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55319 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65119 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11333 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23094 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10918764 - -
          Affymetrix Rat Gene 1.0 ST Array 10918767 - -
          Affymetrix Rat Gene 1.0 ST Array 10918762 - -
          Affymetrix Rat Gene 1.0 ST Array 10918766 - -
          Affymetrix Rat Gene 1.0 ST Array 10918768 - -
          Affymetrix Rat Gene 1.0 ST Array 10918769 - -
          Affymetrix Rat Gene 1.0 ST Array 10918763 - -
          Affymetrix Rat Gene 1.0 ST Array 10918765 - -
          Affymetrix Rat Gene 1.1 ST Array 10918761 - -
          Rat Genome 230 2.0 Array 1369377_at - -
          Rat Genome 230 2.0 Array 1369377_PM_at - -
          Affymetrix HT Rat Focus Array 1369377_at - -
          Affymetrix HT Rat Focus Array 1369377_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800023700.rn.1 TC0800002952.rn.1 RTA TC
          Rat Expression Set 230 1369377_PM_at - -
          Rat Expression Set 230 1369377_at - -
          Affymetrix Rat Gene 1.0 ST Array 10918761 - -
          Rat Genome U34 Set AF041246_at - -
          Rat Genome U34 Set AF041246_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20839140 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357940 - -
          RatRef-12 Expression BeadChip ILMN_66388 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline