Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565387_m1
          See other GNAI3 GE Assays ›
          Gene Symbol:
          Gnai3
          Gene Name:
          guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 3
          Gene Aliases:
          GIP3A
          Chromosome Location:
          Chr.2: 210896779 - 210934733 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565387_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnai3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013106.1 NP_037238.1 3 - 4 392 78
          GenBank mRNA DQ120473.1 - 3 - 4 285 78
          DQ120474.1 - 3 - 4 285 78
          FQ222046.1 - 3 - 4 455 78
          J03219.1 - 3 - 4 401 78
          M20713.1 - 3 - 4 392 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004459.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnai3

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          regulation of heart contraction
          metabolic process
          negative regulation of secretion
          cell division
          GTPase activity
          signal transducer activity
          protein binding
          GTP binding
          protein domain specific binding
          G-protein beta/gamma-subunit complex binding
          G-protein coupled serotonin receptor binding
          GTPase activating protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P056669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171584 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96957 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056669 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056669 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P056669 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019639 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700529452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55262 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93899 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57573 - -
          Agilent Rat cDNA Microarray (G4105A) 700529452 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825967 - -
          Affymetrix Rat Gene 1.0 ST Array 10825972 - -
          Affymetrix Rat Gene 1.0 ST Array 10825964 - -
          Affymetrix Rat Gene 1.0 ST Array 10825970 - -
          Affymetrix Rat Gene 1.0 ST Array 10825969 - -
          Affymetrix Rat Gene 1.0 ST Array 10825968 - -
          Affymetrix Rat Gene 1.0 ST Array 10825971 - -
          Affymetrix Rat Gene 1.0 ST Array 10825963 - -
          Affymetrix Rat Gene 1.0 ST Array 10825965 - -
          Affymetrix Rat Gene 1.0 ST Array 10825966 - -
          Affymetrix Rat Gene 1.1 ST Array 10825962 - -
          Rat Genome 230 2.0 Array 1368029_PM_at - -
          Rat Genome 230 2.0 Array 1368030_at - -
          Rat Genome 230 2.0 Array 1368029_at - -
          Rat Genome 230 2.0 Array 1368030_PM_at - -
          Affymetrix HT Rat Focus Array 1368029_at - -
          Affymetrix HT Rat Focus Array 1368030_at - -
          Affymetrix HT Rat Focus Array 1368029_PM_at - -
          Affymetrix HT Rat Focus Array 1368030_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200030961.rn.1 TC0200004459.rn.1 RTA TC
          Rat Expression Set 230 1368030_at - -
          Rat Expression Set 230 1368029_PM_at - -
          Rat Expression Set 230 1368029_at - -
          Rat Expression Set 230 1368030_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10825962 - -
          Rat Genome U34 Set rc_AI228247_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22309076 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364579 - -
          RatRef-12 Expression BeadChip ILMN_66649 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0