Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00565416_m1
          See other SELP GE Assays ›
          Gene Symbol:
          Selp
          Gene Name:
          selectin P
          Gene Aliases:
          PSELECT
          Chromosome Location:
          Chr.13: 82428943 - 82464629 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00565416_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Selp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013114.1 NP_037246.1 4 - 5 799 97
          XM_006250150.2 XP_006250212.1 5 - 6 913 97
          GenBank mRNA BC101919.1 - 4 - 5 789 7935390 97
          L23088.1 - 4 - 5 799 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000651.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Selp

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte migration
          regulation of cellular extravasation
          inflammatory response
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          positive regulation of platelet activation
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          single organismal cell-cell adhesion
          response to lipopolysaccharide
          regulation of integrin activation
          positive regulation of cell adhesion
          blood vessel morphogenesis
          leukocyte migration
          leukocyte tethering or rolling
          lipopolysaccharide binding
          glycoprotein binding
          heparin binding
          carbohydrate binding
          sialic acid binding
          fucose binding
          calcium-dependent protein binding
          oligosaccharide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11721 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P166514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11721 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166514 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11721 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P166514 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P166514 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11721 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11721 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701437896 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8419 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71426 - -
          Agilent Rat cDNA Microarray (G4105A) 701437896 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765206 - -
          Affymetrix Rat Gene 1.0 ST Array 10765203 - -
          Affymetrix Rat Gene 1.0 ST Array 10765199 - -
          Affymetrix Rat Gene 1.0 ST Array 10765207 - -
          Affymetrix Rat Gene 1.0 ST Array 10765209 - -
          Affymetrix Rat Gene 1.0 ST Array 10765196 - -
          Affymetrix Rat Gene 1.0 ST Array 10765197 - -
          Affymetrix Rat Gene 1.0 ST Array 10765198 - -
          Affymetrix Rat Gene 1.0 ST Array 10765210 - -
          Affymetrix Rat Gene 1.0 ST Array 10765204 - -
          Affymetrix Rat Gene 1.0 ST Array 10765208 - -
          Affymetrix Rat Gene 1.0 ST Array 10765201 - -
          Affymetrix Rat Gene 1.0 ST Array 10765205 - -
          Affymetrix Rat Gene 1.0 ST Array 10765211 - -
          Affymetrix Rat Gene 1.0 ST Array 10765202 - -
          Affymetrix Rat Gene 1.1 ST Array 10765195 - -
          Rat Genome 230 2.0 Array 1391946_at - -
          Rat Genome 230 2.0 Array 1369374_at - -
          Rat Genome 230 2.0 Array 1369374_PM_at - -
          Rat Genome 230 2.0 Array 1391946_PM_at - -
          Affymetrix HT Rat Focus Array 1369374_at - -
          Affymetrix HT Rat Focus Array 1369374_PM_at - -
          Affymetrix HT Rat Focus Array 1391946_PM_at - -
          Affymetrix HT Rat Focus Array 1391946_at - -
          Affymetrix Rat Clariom S Assay PSR1300004529.rn.1 TC1300000651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004527.rn.1 TC1300000651.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004528.rn.1 TC1300000651.rn.1 RTA TC
          Rat Expression Set 230 1391946_at - -
          Rat Expression Set 230 1369374_PM_at - -
          Rat Expression Set 230 1391946_PM_at - -
          Rat Expression Set 230 1369374_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765195 - -
          Rat Genome U34 Set L23088_at - -
          Rat Genome U34 Set L23088_at - -
          Rat Genome U34 Set L23088_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21675267 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367054 - -
          RatRef-12 Expression BeadChip ILMN_70252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline